Merck
  • Neutrophil-Related Gene Expression and Low-Density Granulocytes Associated With Disease Activity and Response to Treatment in Antineutrophil Cytoplasmic Antibody-Associated Vasculitis.
Neutrophil-Related Gene Expression and Low-Density Granulocytes Associated With Disease Activity and Response to Treatment in Antineutrophil Cytoplasmic Antibody-Associated Vasculitis.
Arthritis & rheumatology (Hoboken, N.J.) (2015-04-22)
Peter C Grayson, Carmelo Carmona-Rivera, Lijing Xu, Noha Lim, Zhong Gao, Adam L Asare, Ulrich Specks, John H Stone, Philip Seo, Robert F Spiera, Carol A Langford, Gary S Hoffman, Cees G M Kallenberg, E William St Clair, Nadia K Tchao, Steven R Ytterberg, Deborah J Phippard, Peter A Merkel, Mariana J Kaplan, Paul A Monach
ABSTRACT
To discover biomarkers involved in the pathophysiology of antineutrophil cytoplasmic antibody-associated vasculitis (AAV) and to determine whether low-density granulocytes (LDGs) contribute to gene expression signatures in AAV. The source of clinical data and linked biologic specimens was a randomized controlled treatment trial in AAV. RNA sequencing of whole blood from patients with AAV was performed during active disease at the baseline visit and during remission 6 months later. Gene expression was compared between patients who met versus those who did not meet the primary trial outcome of clinical remission at 6 months (responders versus nonresponders). Measurement of neutrophil-related gene expression was confirmed in peripheral blood mononuclear cells (PBMCs) to validate the findings in whole blood. A negative-selection strategy isolated LDGs from PBMC fractions. Differential expression between responders (n = 77) and nonresponders (n = 35) was detected in 2,346 transcripts at the baseline visit (P < 0.05). Unsupervised hierarchical clustering demonstrated a cluster of granulocyte-related genes, including myeloperoxidase (MPO) and proteinase 3 (PR3). A granulocyte multigene composite score was significantly higher in nonresponders than in responders (P < 0.01) and during active disease than during remission (P < 0.01). This signature strongly overlapped an LDG signature identified previously in lupus (false discovery rate by gene set enrichment analysis <0.01). Transcription of PR3 measured in PBMCs was associated with active disease and treatment response (P < 0.01). LDGs isolated from patients with AAV spontaneously formed neutrophil extracellular traps containing PR3 and MPO. In AAV, increased expression of a granulocyte gene signature is associated with disease activity and decreased response to treatment. The source of this signature is likely LDGs, a potentially pathogenic cell type in AAV.
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
2-Propanol, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
2-Propanol, BioReagent, ≥99.5%
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis