Merck
  • PAH occurrence in chalk river systems from the Jura region (France). Pertinence of suspended particulate matter and sediment as matrices for river quality monitoring.
PAH occurrence in chalk river systems from the Jura region (France). Pertinence of suspended particulate matter and sediment as matrices for river quality monitoring.
Environmental science and pollution research international (2015-07-04)
Axelle Chiffre, François Degiorgi, Nadia Morin-Crini, Audrey Bolard, Etienne Chanez, Pierre-Marie Badot
ABSTRACT
This study investigates the variations of polycyclic aromatic hydrocarbon (PAH) levels in surface water, suspended particulate matter (SPM) and sediment upstream and downstream of the discharges of two wastewater treatment plant (WWTP) effluents. Relationships between the levels of PAHs in these different matrices were also investigated. The sum of 16 US EPA PAHs ranged from 73.5 to 728.0 ng L(-1) in surface water and from 85.4 to 313.1 ng L(-1) in effluent. In SPM and sediment, ∑16PAHs ranged from 749.6 to 2,463 μg kg(-1) and from 690.7 μg kg(-1) to 3,625.6 μg kg(-1), respectively. Investigations performed upstream and downstream of both studied WWTPs showed that WWTP discharges may contribute to the overall PAH contaminations in the Loue and the Doubs rivers. Comparison between gammarid populations upstream and downstream of WWTP discharge showed that biota was impacted by the WWTP effluents. When based only on surface water samples, the assessment of freshwater quality did not provide evidence for a marked PAH contamination in either of the rivers studied. However, using SPM and sediment samples, we found PAH contents exceeding sediment quality guidelines. We conclude that sediment and SPM are relevant matrices to assess overall PAH contamination in aquatic ecosystems. Furthermore, we found a positive linear correlation between PAH contents of SPM and sediment, showing that SPM represents an integrating matrix which is able to provide meaningful data about the overall contamination over a given time span.
Sigma-Aldrich
Pyrene, 98%
Sigma-Aldrich
Anthracene, reagent grade, 97%
Sigma-Aldrich
Pyrene, sublimed grade, 99%
Sigma-Aldrich
Anthracene, suitable for scintillation, ≥99.0% (GC)
Sigma-Aldrich
Benzo[k]fluoranthene, for fluorescence, ≥99%
Supelco
Pyrene, analytical standard
Supelco
Indeno[1,2,3-cd]pyrene, analytical standard
Supelco
Anthracene, analytical standard
Sigma-Aldrich
Anthracene, ReagentPlus®, 99%
Sigma-Aldrich
L-Pyroglutamic acid 2-naphthylamide, ≥99% (TLC)
Sigma-Aldrich
Anthracene, sublimed grade, ≥99%
Sigma-Aldrich
Pyrene, puriss. p.a., for fluorescence, ≥99.0% (GC)
Supelco
Benzo[a]pyrene solution, certified reference material, TraceCERT®, 1000 μg/mL in acetone
Supelco
Benzo[a]pyrene solution, certified reference material, TraceCERT®, 200 μg/mL in methylene chloride
Supelco
Fluoranthene, analytical standard
Supelco
Benzo[a]pyrene, analytical standard, for environmental analysis
Sigma-Aldrich
Hexane, anhydrous, 95%
Sigma-Aldrich
Fluoranthene, 98%
Sigma-Aldrich
Perylene, ≥99%
Sigma-Aldrich
Perylene, sublimed grade, ≥99.5%
Sigma-Aldrich
Benzo[a]pyrene, ≥96% (HPLC)
Sigma-Aldrich
Hexane, ReagentPlus®, ≥99%
Sigma-Aldrich
Hexane, Laboratory Reagent, ≥95%
Sigma-Aldrich
Hexane, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99% (GC)