Merck
  • In vitro bioaccessibility of lead in surface dust and implications for human exposure: A comparative study between industrial area and urban district.
In vitro bioaccessibility of lead in surface dust and implications for human exposure: A comparative study between industrial area and urban district.
Journal of hazardous materials (2015-05-11)
Xiangyang Bi, Zhonggen Li, Guangyi Sun, Jinling Liu, Zhixuan Han
ABSTRACT
In this study, ground surface dust samples from two contrasting areas, a former zinc smelting area in Guizhou Province and a common urban district in Wuhan city, Hubei Province, China, were assessed for in vitro Pb bioaccessibility using a physiologically based extraction test (PBET). Extremely elevated concentrations of Pb (220-6348 mg/kg) and other trace metals were observed in the zinc smelting area. While moderate high metal concentrations (79-1544 mg/kg of Pb) in the urban dusts were attributed to various urban activities, coal combustion and traffic emissions. Lead bioaccessibility in the stomach-phase varied from 17.6 to 76.1% and no significant difference was found between industrial and urban dust samples. Compared with the stomach-phase, Pb bioaccessibility in the more alkaline intestinal-phase was considerably lower (1.2-21.8%). A significantly negative correlation was found between dust Ca concentrations and Pb bioaccessibility in the intestinal-phase, suggesting that Ca plays an important role in reducing the bioaccessible Pb in the intestinal-phase. The estimated Pb exposure based on gastric bioaccessible Pb was 13.9 and 1.8 μg/kg day for children living in the industrial and urban areas, respectively, accounting for 85% and 41% of their corresponding total Pb exposure.
Sigma-Aldrich
Nitric-14N acid solution, ~10 N in H2O, 99.99 atom % 14N
Sigma-Aldrich
Rhodium, powder, 99.95% trace metals basis
Rhodium, wire reel, 0.5m, diameter 0.25mm, hard, 99.9%
Rhodium, foil, not light tested, 25x25mm, thickness 0.001mm, permanent polyester support, 99.85%
Rhodium, wire reel, 0.1m, diameter 0.12mm, as drawn, 99.9%
Rhodium, foil, not light tested, 50x50mm, thickness 0.00075mm, permanent polyester support, 99.85%
Rhodium, foil, 25x25mm, thickness 0.125mm, as rolled, 99.9%
Rhodium, wire reel, 0.5m, diameter 0.12mm, as drawn, 99.9%
Rhodium, microfoil, disks, 10mm, thinness 0.50μm, specific density 620μg/cm2, permanent mylar 3.5μm support, 99.99%
Rhodium, foil, light tested, 15x15mm, thickness 0.05mm, as rolled, 99.9%
Rhodium, wire reel, 0.05m, diameter 0.5mm, as drawn, 99.9%
Rhodium, foil, not light tested, 10x10mm, thickness 0.012mm, as rolled, 99.9%
Rhodium, foil, not light tested, 25x25mm, thickness 0.006mm, as rolled, 99.9%
Rhodium, microfoil, disks, 25mm, thinness 0.1μm, specific density 124μg/cm2, permanent mylar 3.5μm support, 99.9%
Rhodium, foil, not light tested, 25x25mm, thickness 0.012mm, as rolled, 99.9%
Rhodium, wire reel, 0.1m, diameter 0.5mm, as drawn, 99.9%
Rhodium, foil, light tested, 25x25mm, thickness 0.05mm, as rolled, 99.9%
Rhodium, foil, not light tested, 25x25mm, thickness 0.0005mm, permanent polyester support, 99.85%
Rhodium, foil, not light tested, 50x50mm, thickness 0.00025mm, permanent polyester support, 99.9%
Rhodium, wire reel, 0.05m, diameter 0.12mm, as drawn, 99.9%
Rhodium, wire reel, 0.2m, diameter 0.12mm, as drawn, 99.9%
Rhodium, foil, 25x25mm, thickness 0.1mm, as rolled, 99.9%
Rhodium, wire reel, 0.1m, diameter 0.25mm, hard, 99.9%
Rhodium, wire reel, 0.2m, diameter 0.25mm, hard, 99.9%
Rhodium, foil, light tested, 25x25mm, thickness 0.05mm, as rolled, x-ray quality, 99.9%
Rhodium, foil, light tested, 25x25mm, thickness 0.075mm, as rolled, 99.9%
Rhodium, wire reel, 0.2m, diameter 0.5mm, as drawn, 99.9%
Rhodium, foil, 25x25mm, thickness 0.15mm, as rolled, 99.9%
Rhodium, wire reel, 0.05m, diameter 0.25mm, hard, 99.9%
Rhodium, foil, not light tested, 50x50mm, thickness 0.006mm, as rolled, 99.9%