Merck
  • A new role for PGA1 in inhibiting hepatitis C virus-IRES-mediated translation by targeting viral translation factors.
A new role for PGA1 in inhibiting hepatitis C virus-IRES-mediated translation by targeting viral translation factors.
Antiviral research (2015-02-11)
Atsuko Tsukimoto, Ryuichi Sugiyama, Makoto Abe, Hironori Nishitsuji, Yuko Shimizu, Kunitada Shimotohno, Gota Kawai, Hiroshi Takaku
ABSTRACT
Previous studies have demonstrated that cyclopentenone prostaglandins (cyPGs) inhibit the replication of a wide variety of DNA and RNA viruses in different mammalian cell types. We investigated a new role for prostaglandin A1 (PGA1) in the inhibition of hepatitis C virus (HCV)-IRES-mediated translation. PGA1 exhibited dose-dependent inhibitory effects on HCV translation in HCV replicon cells. Furthermore, repetitive PGA1 treatment demonstrated the potential to safely induce the suppression of HCV translation. We also validated a new role for PGA1 in the inhibition of HCV-IRES-mediated translation by targeting cellular translation factors, including the small ribosomal subunit (40S) and eukaryotic initiation factors (eIFs). In pull-down assays, biotinylated PGA1 co-precipitated with the entire HCV IRES RNA/eIF3-40S subunit complex. Moreover, the interactions between PGA1 and the elongation factors and ribosomal subunit were dependent upon HCV IRES RNA binding, and the PGA1/HCV IRES RNA/eIF3-40S subunit complex inhibited HCV-IRES-mediated translation. The novel mechanism revealed in this study may aid in the search for more effective anti-HCV drugs.
Sigma-Aldrich
MISSION® esiRNA, targeting human RPS6
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting human PSMC4
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
SAFC
L-Glutamine
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Rps6