Merck
  • Aqueous foams stabilized by temperature-sensitive hairy polymer particles.
Aqueous foams stabilized by temperature-sensitive hairy polymer particles.
Soft matter (2015-09-30)
S Nakayama, S Yusa, Y Nakamura, S Fujii
ABSTRACT
Submicrometer-sized polystyrene (PS) particles carrying poly[2-(diethylamino)ethyl methacrylate] (PDEA) hairs (PDEA-PS particles) were synthesized by free radical dispersion polymerization. The hydrophilicity-hydrophobicity balance of the PDEA could be tuned by varying the temperature at near neutral pH (a lower critical solution temperature of PDEA at pH 6.86, 41 °C) and therefore these sterically-stabilized particles acted as temperature-sensitive stabilizers for aqueous foams. At 25 °C, where the PDEA hairs were hydrated and PDEA-PS particles were colloidally stable in aqueous media, foams were formed which coalesced with time and the size of the bubble increased. At 40 and 45 °C, where the PDEA hairs were partially non-hydrated and PDEA-PS particles were close to flocculation or weakly flocculated, foams were formed and bubble coalescence and size increase speeds were slower than those observed at 25 °C. At and above 50 °C, where the PDEA hairs were non-hydrated and PDEA-PS particles were heavily flocculated in aqueous media, the more stable cream-like foams whose volume was almost the same for 1 week were formed. Scanning electron microscopy studies indicated that the particles mainly adsorbed at the air-water interface as monolayers at 25 °C and as multilayers at and above 40 °C. The foam stability and structure could be controlled by changing the temperature.
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Supelco
Styrene, analytical standard
Sigma-Aldrich
Aluminum oxide, mesoporous, MSU-X (wormhole), average pore size 3.8 nm
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
N,N-Diethylaniline, ≥99%
Sigma-Aldrich
Styrene, ReagentPlus®, contains 4-tert-butylcatechol as stabilizer, ≥99%