Merck
  • Evaluation of three headspace sorptive extraction coatings for the determination of volatile terpenes in honey using gas chromatography-mass spectrometry.
Evaluation of three headspace sorptive extraction coatings for the determination of volatile terpenes in honey using gas chromatography-mass spectrometry.
Journal of chromatography. A (2015-05-11)
J I Cacho, N Campillo, P Viñas, M Hernández-Córdoba
ABSTRACT
Headspace sorptive extraction (HSSE) was used to preconcentrate seven monoterpenes (eucalyptol, linalool, menthol, geraniol, carvacrol, thymol and eugenol) for separation by gas chromatography and mass spectrometry (GC-MS). Three commercially available coatings for the stir bars, namely Polydimethylsiloxane (PDMS), polyacrilate (PA) and Ethylene glycol-silicone (EG-Silicone), were tested, and the influential parameters both in the adsorption and the thermal desorption steps were optimized. PDMS provided the best sensitivity for linalool, geraniol, menthol and eucalyptol, whereas EG-Silicone was best for extracting the phenolic monoterpenes studied. Considering the average obtained slopes from all compounds, PDMS pointed as the best option, and the analytical characteristics for the HSSE-TD-GC-MS method using this coating were obtained. Quantification of the samples was carried out by matrix-matched calibration using a synthetic honey. Detection limits ranged between 0.007 and 0.032 ng g(-1), depending on the compound. Twelve honey samples of different floral origins were analyzed using the HSSE-GC-MS method, the analytes being detected at concentrations up to 64 ng g(-1).
Product Description
Supelco
Eucalyptol (cineole), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Eugenol, natural, ≥98%, FG
Sigma-Aldrich
Eugenol, ≥98%, FCC, FG
Sigma-Aldrich
Sodium chloride solution, BioUltra, ~5 M in H2O
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Eugenol, ReagentPlus®, 99%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture