Merck
  • Amphiphilic Pickering Emulsifiers Based on Mushroom-Type Janus Particles.
Amphiphilic Pickering Emulsifiers Based on Mushroom-Type Janus Particles.
Langmuir : the ACS journal of surfaces and colloids (2015-07-15)
E Passas-Lagos, F Schüth
ABSTRACT
Iron-based mushroom-type Janus particles consisting of a poly(sytrene-co-divinylbenzene) and a silica moiety both with controllable morphologies were successfully synthesized on the gram scale and investigated as surfactants for Pickering emulsions. Two oil-water model systems, namely toluene-water and vegetable oil-water, were stabilized, giving mainly water-in-oil (w/o) emulsions. By varying several parameters, including Janus particle morphologies and the oil-water ratio, fine-tuning of the emulsion systems was possible; it was even possible to invert the continuous phase to an oil-in-water (o/w) system. Furthermore, the emulsions were stable against coalescence and sedimentation and could be easily separated by centrifugation or a strong magnet. The synthesized mushroom-type Janus particles are suitable for creating Pickering emulsions and can be used as building blocks for creating nanostructures with tailored properties for specific applications.
Sigma-Aldrich
Rhodamine 6G, suitable for fluorescence, BioReagent
Sigma-Aldrich
Divinylbenzene, technical grade, 80%
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Divinylbenzene, technical grade, 55%
Sigma-Aldrich
Glycidyl methacrylate, 97%, contains 100 ppm monomethyl ether hydroquinone as inhibitor
Sigma-Aldrich
Rhodamine 6G, Dye content 99 %
Sigma-Aldrich
L-Lysine, crystallized, ≥98.0% (NT)
Sigma-Aldrich
Glycidyl methacrylate, ≥97.0% (GC)
Sigma-Aldrich
Rhodamine 6G, Dye content ~95 %
Sigma-Aldrich
Styrene, ReagentPlus®, contains 4-tert-butylcatechol as stabilizer, ≥99%
Sigma-Aldrich
L-Lysine, ≥98% (TLC)
Sigma-Aldrich
Iron oxide(II,III), magnetic nanoparticles solution, 5 nm avg. part. size (TEM), amine functionalized, 1 mg/mL Fe in H2O, dispersion
Sigma-Aldrich
Iron oxide(II,III), magnetic nanoparticles solution, 10 nm avg. part. size (TEM), PEG functionalized, 1 mg/mL Fe in H2O, dispersion
Sigma-Aldrich
Iron oxide(II,III), magnetic nanoparticles solution, 10 nm avg. part. size (TEM), carboxylic acid functionalized, 5 mg/mL Fe in H2O, dispersion
Sigma-Aldrich
Iron oxide(II,III), magnetic nanoparticles solution, 5 nm avg. part. size (TEM), PEG functionalized, 1 mg/mL Fe in H2O, dispersion
Sigma-Aldrich
Iron oxide(II,III), magnetic nanoparticles solution, 30 nm avg. part. size (TEM), amine functionalized, 1 mg/mL Fe in H2O, dispersion
Sigma-Aldrich
Iron oxide(II,III), magnetic nanoparticles solution, 5 nm avg. part. size, 5 mg/mL in toluene
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Iron oxide(II,III), magnetic nanoparticles solution, 20 nm avg. part. size, 5 mg/mL in toluene
Sigma-Aldrich
Iron oxide(II,III), magnetic nanoparticles solution, 10 nm avg. part. size, 5 mg/mL in toluene
Sigma-Aldrich
Iron oxide(II,III), magnetic nanoparticles solution, 5 nm avg. part. size, 5 mg/mL in H2O
Sigma-Aldrich
Iron oxide(II,III), magnetic nanoparticles solution, 20 nm avg. part. size, 5 mg/mL in H2O
Sigma-Aldrich
Iron oxide(II,III), magnetic nanoparticles solution, 10 nm avg. part. size, 5 mg/mL in H2O
Sigma-Aldrich
Toluene, anhydrous, 99.8%
Sigma-Aldrich
Iron(II,III) oxide, powder, <5 μm, 95%
Sigma-Aldrich
Iron(II,III) oxide, 99.99% trace metals basis
Sigma-Aldrich
2-Propanol, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Ammonium persulfate, BioUltra, for molecular biology, ≥98.0% (RT)
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
Ammonium persulfate, BioXtra, ≥98.0%