Merck
  • The Adh adhesin domain is required for trimeric autotransporter Apa1-mediated Actinobacillus pleuropneumoniae adhesion, autoaggregation, biofilm formation and pathogenicity.
The Adh adhesin domain is required for trimeric autotransporter Apa1-mediated Actinobacillus pleuropneumoniae adhesion, autoaggregation, biofilm formation and pathogenicity.
Veterinary microbiology (2015-03-31)
Lei Wang, Wanhai Qin, Shuxin Yang, Ruidong Zhai, Liang Zhou, Changjiang Sun, Fengguang Pan, Qun Ji, Yu Wang, Jingmin Gu, Xin Feng, Chongtao Du, Wenyu Han, P R Langford, Liancheng Lei
ABSTRACT
Actinobacillus pleuropneumoniae is a causative agent of porcine pleuropneumonia, which is a highly contagious endemic disease of pigs. Adhesion is a critical first step in the infection process. Trimeric autotransporter adhesions (TAAs) have been identified as novel virulence factors; however, little is known on their roles in A. pleuropneumoniae pathogenicity. Here, our data show that YadA-like head region (Adh) of Apa1 was the optimal adhesion functional domain via segment expression and adhesion assays in vitro. Additionally, Adh induced partial protection against A. pleuropneumoniae 5b L20 and serotypes 1, 3, and 5a in mice. The deletion of Adh gene significantly decreased autoaggregation, biofilm formation and adherence to host cells in vitro. Furthermore, with delaying of clinical symptoms, reducing production of pro-inflammatory cytokines and lessening the lung injury after infection, Adh deletion strain (5bϕAdh) significantly reduced the pathogenicity to piglets. To elucidate the mechanism of lung injury, the differentially expressed genes in the lung tissues of piglets infected with the 5b L20 or 5bϕAdh strains were investigated using microarray analysis and validated by qRT-PCR. Compared with the 5b L20 infected piglets, 495 genes were differentially expressed in 5bϕAdh infected lung tissue (221 upregulated and 274 downregulated). Especially, the antigen processing and presentation gene IFI30 was increased following infection with the 5bϕAdh strain. Thus, Adh may enhance pathogenicity by depressing host immune recognition. We conclude that the head domain of the A. pleuropneumoniae trimeric autotransporter Apa1 regulates autoagglutination, biofilm formation, adhesion to host cells and pathogenicity.
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine
SAFC
L-Glutamine
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide, pkg of 10 mg (per vial)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide, pkg of 20 mg (per vial)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide, pkg of 50 mg (per vial)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, ≥96.5% (HPLC), ≥96.5% (spectrophotometric assay), from yeast
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide lithium salt from Saccharomyces cerevisiae, ≥95%
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, suitable for cell culture, ≥96.5% (HPLC), ≥96.5% (spectrophotometric assay), from yeast
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, Grade AA-1
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, ≥98%, BioUltra, from yeast
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, ≥99%
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, purified by column chromatography, ≥99%
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, ≥95% (HPLC)