Merck
  • Enhancement of nutritionally significant constituents of black currant seeds by chemical elicitor application.
Enhancement of nutritionally significant constituents of black currant seeds by chemical elicitor application.
Food chemistry (2015-10-17)
Gema Flores, María Luisa Ruiz del Castillo
ABSTRACT
Black currant seeds are obtained as a residue during juice production. Black currant seed oil contains high amounts of nutritionally desirable constituents such as γ-linolenic acid (GLA), α-linolenic acid (ALA) and stearidonic acid (SA), as well as certain phenolic acids, which act as natural antioxidants. Fatty acids and phenolic acids of seeds from black currant cultivars after elicitation with methyl jasmonate (MJ) were examined. GLA contents around 25% with respect to total fatty acid content were measured in seeds after pre-harvest treatment of black currants with 0.02mM MJ in 0.05% Tween-20. High GLA samples also exhibited high SA content (higher than 10% with respect to total fatty acid content); however, ALA dropped (from 16% to 10%). High GLA content seeds also showed increased contents of gallic, caffeic, p-coumaric and ferulic acids. In particular, seeds from 0.02mM MJ treated Ben Hope black currants exerted contents of gallic, caffeic, p-coumaric and ferulic acids of 201.4, 125.9, 201.3 and 112.5μgg(-1)vs 124.3, 58.6, 165.4 and 95.8μgg(-1) measured in seeds from untreated Ben Hope black currants. Comparable results were obtained for Ben Alder and Ben Gairn berries. Chemical elicitation with 0.02 MJ is proposed as an industrial practice in such a way that, after consideration of quality issues, it would be obtained high added value black currant seeds.
Sigma-Aldrich
Sodium methoxide, reagent grade, 95%, powder
Sigma-Aldrich
trans-Ferulic acid, ≥99%
Sigma-Aldrich
Sodium methoxide solution, ACS reagent, 0.5 M CH3ONa in methanol (0.5N)
Sigma-Aldrich
trans-Ferulic acid, 99%
Sigma-Aldrich
Sodium methoxide solution, 25 wt. % in methanol
Sigma-Aldrich
Butylated hydroxytoluene, ≥99%, FCC, FG