Merck
  • ERK and RSK are necessary for TRH-induced inhibition of r-ERG potassium currents in rat pituitary GH3 cells.
ERK and RSK are necessary for TRH-induced inhibition of r-ERG potassium currents in rat pituitary GH3 cells.
Cellular signalling (2015-05-30)
Luis Carretero, Pablo Llavona, Alejandro López-Hernández, Pedro Casado, Pedro R Cutillas, Pilar de la Peña, Francisco Barros, Pedro Domínguez
ABSTRACT
The transduction pathway mediating the inhibitory effect that TRH exerts on r-ERG channels has been thoroughly studied in GH3 rat pituitary cells but some elements have yet to be discovered, including those involved in a phosphorylation event(s). Using a quantitative phosphoproteomic approach we studied the changes in phosphorylation caused by treatment with 1μM TRH for 5min in GH3 cells. The activating residues of Erk2 and Erk1 undergo phosphorylation increases of 5.26 and 4.87 fold, respectively, in agreement with previous reports of ERK activation by TRH in GH3 cells. Thus, we studied the possible involvement of ERK pathway in the signal transduction from TRH receptor to r-ERG channels. The MEK inhibitor U0126 at 0.5μM caused no major blockade of the basal r-ERG current, but impaired the TRH inhibitory effect on r-ERG. Indeed, the TRH effect on r-ERG was also reduced when GH3 cells were transfected with siRNAs against either Erk1 or Erk2. Using antibodies, we found that TRH treatment also causes activating phosphorylation of Rsk. The TRH effect on r-ERG current was also impaired when cells were transfected with any of two different siRNAs mixtures against Rsk1. However, treatment of GH3 cells with 20nM EGF for 5min, which causes ERK and RSK activation, had no effect on the r-ERG currents. Therefore, we conclude that in the native GH3 cell system, ERK and RSK are involved in the pathway linking TRH receptor to r-ERG channel inhibition, but additional components must participate to cause such inhibition.
Sigma-Aldrich
2-Butanone, FCC, FG
Sigma-Aldrich
PD184352, ≥98% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting human RPS6KA1
Sigma-Aldrich
MISSION® esiRNA, targeting human RPS6KA2
Sigma-Aldrich
E-4031, ≥98% (HPLC), lyophilized powder
Sigma-Aldrich
MISSION® esiRNA, targeting human RPS6KA3
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Rps6ka3
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Rps6ka1
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Rps6ka2