Merck
Apaf1 inhibition promotes cell recovery from apoptosis.
Protein & cell (2015-09-13)
Anna Gortat, Mónica Sancho, Laura Mondragón, Àngel Messeguer, Enrique Pérez-Payá, Mar Orzáez
ABSTRACT
The protein apoptotic protease activating factor 1 (Apaf1) is the central component of the apoptosome, a multiprotein complex that activates procaspase-9 after cytochrome c release from the mitochondria in the intrinsic pathway of apoptosis. We have developed a vital method that allows fluorescence-activated cell sorting of cells at different stages of the apoptotic pathway and demonstrated that upon pharmacological inhibition of Apaf1, cells recover from doxorubicin- or hypoxia-induced early apoptosis to normal healthy cell. Inhibiting Apaf1 not only prevents procaspase-9 activation but delays massive mitochondrial damage allowing cell recovery.
Sigma-Aldrich
MISSION® esiRNA, targeting human APAF1
Sigma-Aldrich
MISSION® esiRNA, targeting human BECN1
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Apaf1
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Sodium phosphate, 96%
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Becn1
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology