Merck
  • Sensitive and validated LC-MS/MS methods to evaluate mycophenolic acid pharmacokinetics and pharmacodynamics in hematopoietic stem cell transplant patients.
Sensitive and validated LC-MS/MS methods to evaluate mycophenolic acid pharmacokinetics and pharmacodynamics in hematopoietic stem cell transplant patients.
Biomedical chromatography : BMC (2015-01-28)
Misaki Kawanishi, Ikuko Yano, Kazuaki Yoshimura, Takashi Yamamoto, Sachiyo Hashi, Satohiro Masuda, Tadakazu Kondo, Akifumi Takaori-Kondo, Kazuo Matsubara
ABSTRACT
Monitoring of pharmacodynamics in addition to pharmacokinetics is one of strategies to individualize mycophenolate mofetil therapy. The purpose of this study was to develop sensitive liquid chromatography-tandem mass spectrometry (LC-MS/MS) methods for evaluation of the pharmacokinetics and pharmacodynamics of mycophenolic acid (MPA). Concentrations of mycophenolic acid glucuronide (MPAG), mycophenolic acid acyl-glucuronide, as well as unbound MPA and MPAG, were determined, and inosine-5'-monophosphate dehydrogenase activity was calculated by measuring concentrations of produced xanthosine-5'-monophosphate (XMP) and intracellular adenosine-5'-monophosphate after incubation of peripheral blood mononuclear cell (PBMC) lysates. A metal-free Mastro(TM) column and two gradient patterns were used to improve the quantification limit of XMP to 0.1 μM. In the clinical MPA concentration range, the linearity of the calibration curve, inter- and intra-day precision and accuracy satisfied the relevant US Food and Drug Administration guidelines. The MPA concentrations in hematopoietic stem cell transplant (HSCT) patients determined by the enzyme assay and the present LC-MS/MS method showed a good correlation (r(2) = 0.95, p < 0.001). In this study, we report sensitive and validated LC-MS/MS methods to evaluate the pharmacokinetics and pharmacodynamics of MPA, which are sufficiently sensitive to assess small quantities of PBMC lysates collected shortly after HSCT.
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, ≥98%, BioUltra, from yeast
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, suitable for cell culture, ≥96.5% (HPLC), ≥96.5% (spectrophotometric assay), from yeast
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, purified by column chromatography, ≥99%
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, ≥96.5% (HPLC), ≥96.5% (spectrophotometric assay), from yeast
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide lithium salt from Saccharomyces cerevisiae, ≥95%
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, ≥99%
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, ≥95% (HPLC)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, Grade AA-1
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide, pkg of 20 mg (per vial)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide, pkg of 10 mg (per vial)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide, pkg of 50 mg (per vial)
Sigma-Aldrich
Adenosine 5′-monophosphate disodium salt, ≥99.0% (HPLC)
Sigma-Aldrich
Propylene glycol monomethyl ether acetate, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Adenosine 5′-monophosphate sodium salt, from yeast, ≥99%