Merck
Lead exposure causes thyroid abnormalities in diabetic rats.
International journal of clinical and experimental medicine (2015-07-30)
Salah Al Zadjali, Abderrahim Nemmar, Mohamed Abdelmonem Ay Fahim, Sheikh Azimullah, Dhanasekaran Subramanian, Javed Yasin, Naheed Amir, Mohammed Yousif Hasan, Abdu Adem
ABSTRACT
Lead is a widely-spread environmental pollutant and a commonly-used industrial chemical that can cause multisystemic adverse health effects. However, the effects of lead exposure on diabetic animals have not been reported so far. The aim of this study is to evaluate the effects of lead exposure on thyroid, renal and oxidative stress markers in diabetic Wistar rats. Diabetes was induced with an intraperitoneal (i.p.) injection of streptozocin (STZ). Six weeks later, rats were exposed i.p. to either distilled water (control group) or 25, 50 and 100 mg/kg of lead acetate (treatment groups). We found a positive relationship between the administered doses of lead acetate and its measured levels in blood samples (P < 0.01). Treatment of diabetic animals with lead acetate resulted in significant weight loss (P < 0.001). It also caused an increase in thyroid stimulating hormone levels (P < 0.05) and reductions in thyroxine (P < 0.05) and triiodothyronine levels (P < 0.01), a clinical picture consistent with hypothyroidism. Lead acetate exposure increased urea levels (P < 0.05) and caused a significant decrease in creatinine (P < 0.05). Besides, while the concentrations of malondialdehyde were not affected, glutathione stores were depleted (P < 0.01); in response to lead exposure. In conclusion, exposure of diabetic rats to lead acetate resulted in weight loss, clinical hypothyroidism, renal damage and oxidative stress.
Sigma-Aldrich
Rhodium(III) nitrate solution, ~10 % (w/w) (Rh in >5 wt. % HNO3)
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, for molecular biology, 0.5 M in H2O, DNase, RNase, NICKase and protease, none detected
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, reagent grade, 98.5-101.5% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, ACS reagent, 99.0-101.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, 98.5-101.5%, BioUltra
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, suitable for electrophoresis, for molecular biology, 99.0-101.0% (titration)
Sigma-Aldrich
Rhodium, powder, 99.95% trace metals basis
Rhodium, wire reel, 0.5m, diameter 0.25mm, hard, 99.9%
Rhodium, foil, not light tested, 25x25mm, thickness 0.001mm, permanent polyester support, 99.85%
Rhodium, wire reel, 0.1m, diameter 0.12mm, as drawn, 99.9%
Rhodium, foil, not light tested, 50x50mm, thickness 0.00075mm, permanent polyester support, 99.85%
Rhodium, foil, 25x25mm, thickness 0.125mm, as rolled, 99.9%
Rhodium, wire reel, 0.5m, diameter 0.12mm, as drawn, 99.9%
Rhodium, microfoil, disks, 10mm, thinness 0.50μm, specific density 620μg/cm2, permanent mylar 3.5μm support, 99.99%
Rhodium, foil, light tested, 15x15mm, thickness 0.05mm, as rolled, 99.9%
Rhodium, wire reel, 0.05m, diameter 0.5mm, as drawn, 99.9%
Rhodium, foil, not light tested, 10x10mm, thickness 0.012mm, as rolled, 99.9%
Rhodium, foil, not light tested, 25x25mm, thickness 0.006mm, as rolled, 99.9%
Rhodium, microfoil, disks, 25mm, thinness 0.1μm, specific density 124μg/cm2, permanent mylar 3.5μm support, 99.9%
Rhodium, foil, not light tested, 25x25mm, thickness 0.012mm, as rolled, 99.9%
Rhodium, wire reel, 0.1m, diameter 0.5mm, as drawn, 99.9%
Rhodium, foil, light tested, 25x25mm, thickness 0.05mm, as rolled, 99.9%
Rhodium, foil, not light tested, 25x25mm, thickness 0.0005mm, permanent polyester support, 99.85%
Rhodium, foil, not light tested, 50x50mm, thickness 0.00025mm, permanent polyester support, 99.9%
Rhodium, wire reel, 0.05m, diameter 0.12mm, as drawn, 99.9%
Rhodium, wire reel, 0.2m, diameter 0.12mm, as drawn, 99.9%