Merck
  • First N-Heterocyclic Carbenes Relying on the Triazolone Structural Motif: Syntheses, Modifications and Reactivity.
First N-Heterocyclic Carbenes Relying on the Triazolone Structural Motif: Syntheses, Modifications and Reactivity.
Chemistry (Weinheim an der Bergstrasse, Germany) (2015-09-24)
Markus Jonek, Janina Diekmann, Christian Ganter
ABSTRACT
4-Phenylsemicarbazide and 1,5-diphenylcarbazide are suitable starting materials for the syntheses of N-heterocyclic carbene (NHC) compounds with new backbone structures. In the first case, cyclisation and subsequent methylation leads to a cationic precursor whose deprotonation affords the triazolon-ylidene 2, which was converted to the corresponding sulfur and selenium adducts and a range of metal complexes. In contrast, cyclisation of diphenylcarbazide affords a neutral betain-type NHC-precursor 7, which is not in equilibrium with its carbene tautomer 7a. Precursor 7 can either be deprotonated to give the anionic NHC 8 or methylated at the N or O atom of the backbone resulting in two isomeric cationic species 16 and 20. Deprotonation of the latter two provides neutral NHC compounds with a carboxamide or carboximidate backbone, respectively. The ligand properties of the new NHC compounds were evaluated by IR and (77) Se NMR spectroscopy. Tolman electronic parameter (TEP) values range from 2050 to 2063 cm(-1) with the anionic NHC 8 being the best overall donor.
Sigma-Aldrich
Tetrahydrofuran, anhydrous, ≥99.9%, inhibitor-free
Supelco
Tetrahydrofuran, HPLC grade, ≥99.9%, inhibitor-free
Sigma-Aldrich
Tetrahydrofuran, anhydrous, contains 250 ppm BHT as inhibitor, ≥99.9%
Sigma-Aldrich
Tetrahydrofuran, ACS reagent, ≥99.0%, contains 250 ppm BHT as inhibitor
Sigma-Aldrich
Tetrahydrofuran, ReagentPlus®, ≥99.0%, contains 250 ppm BHT as inhibitor
Sigma-Aldrich
Tetrahydrofuran, contains 250 ppm BHT as inhibitor, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.9%
Sigma-Aldrich
Tetrahydrofuran, contains 250 ppm BHT as inhibitor, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium bis(trimethylsilyl)amide solution, 0.6 M in toluene
Sigma-Aldrich
Sodium bis(trimethylsilyl)amide solution, 40% in THF
Sigma-Aldrich
Sodium bis(trimethylsilyl)amide, 95%
Sigma-Aldrich
Potassium tert-butoxide solution, 1.0 M in THF
Sigma-Aldrich
Sodium bis(trimethylsilyl)amide solution, 1.0 M in THF
Sigma-Aldrich
Potassium tert-butoxide, sublimed grade, 99.99% trace metals basis
Sigma-Aldrich
Potassium tert-butoxide, reagent grade, ≥98%