Merck
  • The blood pressure effect and related plasma levels of flavan-3-ols in spontaneously hypertensive rats.
The blood pressure effect and related plasma levels of flavan-3-ols in spontaneously hypertensive rats.
Food & function (2015-08-22)
Mar Quiñones, Maria Margalef, Anna Arola-Arnal, Begoña Muguerza, Marta Miguel, Amaya Aleixandre
ABSTRACT
We studied the short-term antihypertensive effect of flavan-3-ols (-)-epicatechin, (+)-catechin and (-)-catechin, in spontaneously hypertensive rats (SHR). Plasma metabolites and the corresponding plasma antioxidant capacity were determined. All the assayed flavan-3-ols decreased systolic blood pressure (SBP) in SHR. Their antihypertensive effects were less pronounced than that of Captopril (50 mg kg(-1)) and were not shown in normotensive Wistar-Kyoto rats. 6 mg kg(-1) (-)-epicatechin caused the maximum decrease in SBP. The maximum effects of the catechin monomers were observed post-administration of 0.5 mg kg(-1) of flavan-3-ols, (-)-catechin being the least effective among the three assayed compounds. Glucuronide and methyl glucuronide metabolites were obtained in the flavan-3-ol treated SHR, but it was not possible to relate the antihypertensive effect of the assayed flavan-3-ols with a concrete plasma metabolite or with their antioxidant effect. In conclusion, the studied flavan-3-ols could be responsible for the antihypertensive effect of cocoa products.
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Pyrocatechol, purified by sublimation, ≥99.5%
Sigma-Aldrich
(−)-Epicatechin, ≥98% (HPLC), from green tea
Sigma-Aldrich
(−)-Epicatechin, ≥90% (HPLC)
Sigma-Aldrich
Pyrocatechol, ≥99%
Sigma-Aldrich
(−)-Catechin, ≥97% (HPLC), from green tea
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Acetone, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetone, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Captopril, meets USP testing specifications
Sigma-Aldrich
Captopril, ≥98% (HPLC), powder
Sigma-Aldrich
1,2-Dihydroxybenzene, ReagentPlus®, ≥99%
Sigma-Aldrich
Fluorescein, for fluorescence, free acid
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%