Merck
Flexible Lamination Encapsulation.
Advanced materials (Deerfield Beach, Fla.) (2015-06-24)
Min-Ho Park, Jin-You Kim, Tae-Hee Han, Tae-Sik Kim, Hobeom Kim, Tae-Woo Lee
ABSTRACT
A novel flexible encapsulation method (Flex Lami-capsulation) is reported, which can be applied in the roll-to-roll process for mass production of organic electronic devices. Flex Lami-capsulation is very simple, fast, and getter-free, and is as effective as glass encapsulation. Use of this method is feasible in large-area flexible displays and does not have the drawbacks of conventional encapsulation methods.
Sigma-Aldrich
D.E.R. 332, used as embedding medium
Tantalum(V) ethoxide, packaged for use in deposition systems
Sigma-Aldrich
Tantalum(V) ethoxide, 99.98% trace metals basis