Merck
  • Cape Gooseberry [Physalis peruviana L.] Calyces Ameliorate TNBS Acid-induced Colitis in Rats.
Cape Gooseberry [Physalis peruviana L.] Calyces Ameliorate TNBS Acid-induced Colitis in Rats.
Journal of Crohn's & colitis (2015-07-30)
Jenny Castro, Yanet Ocampo, Luis Franco
ABSTRACT
Physalis peruviana [cape gooseberry] is highly appreciated for its commercial value. The Colombian ecotype is in great demand in the international market, particularly for the unique morphological characteristics of the calyx, which has extended use as a traditional herbal remedy in Colombia because of its anti-inflammatory properties. In this work, the anti-inflammatory activity of the total ethereal extract of Physalis peruviana calyces was evaluated in preventive and therapeutic protocols in a TNBS acid-induced colitis rat model. Colitis was induced by intrarectal administration of TNBS. An evaluation of macroscopic and histopathological parameters in colonic tissue was performed, along with the determination of myeloperoxidase enzyme activity, cytokine levels and gene expression. Additionally, effects on nitric oxide release by lipopolysaccharide [LPS]-stimulated RAW264.7 macrophages and the scavenging activity of DPPH and ABTS free radicals were determined. The treatment with the Physalis peruviana extract produced a significant improvement in the colonic tissue at both macroscopic and histological levels. IL-1β and TNF-α production was reduced by the extract in both experimental approaches. The groups treated with Physalis peruviana showed a tendency to MUC2 up-regulation and down-regulation of COX-2, iNOS, NLRP3, IL-1β, IL-6 and IL-10 expression. Nitric oxide release in RAW264.7 macrophages was significantly inhibited. The Physalis peruviana extract showed intestinal anti-inflammatory activity in the TNBS-induced colitis model, placing this species' calyx, a natural derivative, as a promising source of metabolites that could be used in treatment for inflammatory bowel disease.
Sigma-Aldrich
Sulfanilamide, puriss. p.a., ≥98% (calc. to the dried substance)
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Trypan Blue, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Sulfanilamide, ≥98%
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Trypan Blue, Dye content 60 %, ≥80% (HPLC)
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Sigma-Aldrich
Zirconyl chloride octahydrate, reagent grade, 98%
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Potassium persulfate, 99.99% trace metals basis
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
1,1-Diphenyl-2-picrylhydrazine, 97%
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Sigma-Aldrich
2,2-Diphenyl-1-picrylhydrazyl
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Picrylsulfonic acid solution, 5 % (w/v) in H2O, BioReagent, suitable for determination of primary amines
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%