Merck
Antitumor activity of a novel survivin siRNA.
Pakistan journal of pharmaceutical sciences (2015-11-04)
Yuhuan Li, Yujing Li, Robert J Lee, Xi Wang, Lin Du, Lingzhi Zhao, Yan Liu, Lesheng Teng
ABSTRACT
Breast cancer resistance to therapy can result from expression of antiapoptotic genes. Survivin is an antiapoptotic gene that is over expressed in most human tumors. RNA interference using short interfering RNA (siRNA) can be used to specifically inhibit survivin expression. A novel siRNA targeting survivin was used to process MCF-7 cells. Cellular survivin mRNA and protein levels were determined by real-time qRT-PCR and Western blot, respectively. Cellular morphology and cell cycle were determined by fluorescence microscopy and flow cytometry. Cell proliferation was measured by MTT assay. Our data showed that the novel survivin-targeted siRNA could efficiently knockdown the expression of survivin, inhibit cell proliferation and cell cycle, especially at the G2/M checkpoint. These data suggest that the siRNA has potential for therapeutic applications.
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Birc5