Merck
  • Rapid and Versatile Photonic Annealing of Graphene Inks for Flexible Printed Electronics.
Rapid and Versatile Photonic Annealing of Graphene Inks for Flexible Printed Electronics.
Advanced materials (Deerfield Beach, Fla.) (2015-10-01)
Ethan B Secor, Bok Y Ahn, Theodore Z Gao, Jennifer A Lewis, Mark C Hersam
ABSTRACT
Intense pulsed light (IPL) annealing of graphene inks is demonstrated for rapid post-processing of inkjet-printed patterns on various substrates. A conductivity of ≈25,000 S m(-1) is achieved following a single printing pass using a concentrated ink containing 20 mg mL(-1) graphene, establishing this strategy as a practical and effective approach for the versatile and high-performance integration of graphene in printed and flexible electronics.
Sigma-Aldrich
Carbon nanofibers, graphitized, platelets(conical), >98% carbon basis, D × L 100 nm × 20-200 μm
Sigma-Aldrich
Carbon nanofibers, pyrolitically stripped, platelets(conical), >98% carbon basis, D × L 100 nm × 20-200 μm
Sigma-Aldrich
Terpineol, mixture of isomers
Sigma-Aldrich
Cyclohexanone, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Cyclohexanone, 99.8%
Sigma-Aldrich
Toluene, anhydrous, 99.8%
Sigma-Aldrich
Methane-12C, 13C-depleted, 99.9 atom % 12C
Sigma-Aldrich
Cyclohexanone, ReagentPlus®, 99.8%
Sigma-Aldrich
Cyclohexanone, ACS reagent, ≥99.0%
Sigma-Aldrich
N,O-Bis(trimethylsilyl)acetamide, synthesis grade, ≥95%
Sigma-Aldrich
Graphene ink, for inkjet printing, photonically annealable
Sigma-Aldrich
Graphene ink for spin/spray coating photonically annealable, for spin-coating, spray-coating, photonic annealing
Sigma-Aldrich
Graphene ink, for inkjet printing, with ethyl cellulose in cyclohexanone and terpineol, inkjet printable
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, Laboratory Reagent, ≥99.3%
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Carbon - Vitreous, foil, 50x50mm, thickness 4.0mm, glassy carbon
Carbon - Vitreous, rod, 200mm, diameter 1.0mm, glassy carbon
Carbon - Vitreous, foil, 100x100mm, thickness 2.0mm, glassy carbon
Carbon - Vitreous, foil, 8x8mm, thickness 0.5mm, glassy carbon
Carbon - Vitreous, foil, 100x100mm, thickness 1.0mm, glassy carbon
Carbon - Vitreous, foil, 100x100mm, thickness 6.0mm, glassy carbon
Carbon - Vitreous, rod, 200mm, diameter 10mm, glassy carbon
Carbon - Vitreous, foam, 150x150mm, thickness 2.5mm, bulk density 0.05g/cm3, porosity 96.5%
Carbon - Vitreous, rod, 200mm, diameter 3.0mm, glassy carbon
Carbon - Vitreous, foil, 10x10mm, thickness 1.0mm, glassy carbon
Carbon - Vitreous, foam, 150x150mm, thickness 3.2mm, bulk density 0.05g/cm3, porosity 96.5%
Carbon - Vitreous, foam, 150x150mm, 0.05g.cmué, porosity 96.5%, 24 pores/cm