Merck
  • Isolation and characterization of meniscus derived stem cells from rabbit as a possible treatment for damaged meniscus.
Isolation and characterization of meniscus derived stem cells from rabbit as a possible treatment for damaged meniscus.
Current stem cell research & therapy (2015-05-20)
Jianchao Gui, Jianying Zhang, He Huang
ABSTRACT
The successful surgical restoration of damaged meniscus has been a challenge, largely owing to a lack of characterization of meniscus cells and their precursors. Numerous strategies to repair or replace meniscus have achieved only limited success. Several recent studies have shown beneficial effect of mesenchymal stem cells in meniscus repair. The objective of our study was to characterize meniscus derived mesenchymal stem cells in terms of, colony formation, proliferation, multi potency and self-renewal capacity. Mesenchymal stem cells were isolated from menisci, patellar tendon and bone marrow of rabbits. The multi differentiating potential, colony, morphology and proliferation were studied in vitro. The expression of differential markers was studied by immunocytochemistry, qPCR and western blotting. Three groups of cells appeared similar in colony formation and morphology. All of them were found to express high levels of stem cell markers including SSEA-4, Nanog and nucleostemin. High level of collagen II expression was detected in meniscus derived stem cells. Moreover, these cells appeared to have a pronounced tendency to chondrogenic differentiation under specialized culture conditions. Meniscus-derived mesenchymal stem cells (MMSCs) possessed all the necessary criteria of stem cells, including clonogenicity, self-renewal and multipotent differentiation capacity and possessed a tendency to differentiate into chondrocytes. Our results offer new insights into the biology of meniscus cells, and may assist in future strategies to treat damaged meniscus.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting human CEACAM1
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
2-Mercaptoethanol, for molecular biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Selenous acid, 98%
Sigma-Aldrich
Selenous acid, 99.999% trace metals basis
Sigma-Aldrich
Safranin O, for microscopy (Bact., Bot., Hist.), indicator (pH 0.3-1.0)
Sigma-Aldrich
bisBenzimide H 33342 trihydrochloride, ≥98% (HPLC and TLC)
Sigma-Aldrich
Safranin O, Dye content ≥85 %