Merck
  • Surface plasmon resonance-based immunoassay for human C-reactive protein.
Surface plasmon resonance-based immunoassay for human C-reactive protein.
The Analyst (2015-05-13)
S K Vashist, E M Schneider, J H T Luong
ABSTRACT
A rapid and highly-sensitive surface plasmon resonance (SPR)-based immunoassay (IA) has been developed and validated for detecting human C-reactive protein (CRP), a specific biomarker for inflammatory and metabolic disorders, and infections. The 1-ethyl-3-(3-dimethylaminopropyl)-carbodiimide hydrochloride (EDC)-activated protein A/G (Pr A/G) was diluted in 1% (v/v) 3-aminopropyltriethoxysilane (APTES), dispensed on a KOH-treated gold (Au)-coated SPR chip, and incubated for 30 min. The Pr A/G functionalized Au SPR chip was then bound to anti-human CRP capture antibody (Ab), blocked with bovine serum albumin, and subsequently used for the detection of CRP. The highly-simplified oriented Ab immobilization strategy enabled the leach-proof binding of capture Ab in 5-fold shorter time than conventional procedures. The developed IA detected 1.2-80 ng mL(-1) of CRP with a limit of detection (LOD) and a limit of quantification (LOQ) of 1.2 ng mL(-1) and 4.6 ng mL(-1), respectively. It detected CRP spiked in diluted human whole blood, serum and plasma as well as the CRP levels in the ethylenediaminetetraacetic acid (EDTA) plasma samples of patients with the same precision as the clinically-accredited analyzer-based IA and conventional CRP sandwich ELISA. The Ab-bound SPR chips stored at 4 °C retained their functional activity for 10 weeks, resulting in significant reduction in the overall analysis time.
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, ≥99.5% (titration)
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
SAFC
HEPES
SAFC
HEPES
Sigma-Aldrich
Sodium chloride, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
HEPES, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, BioUltra, ~5 M in H2O
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, packaged for use in deposition systems, ≥98%
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, ≥98.0%
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder