Merck
  • A novel approach for LC-MS/MS-based chiral metabolomics fingerprinting and chiral metabolomics extraction using a pair of enantiomers of chiral derivatization reagents.
A novel approach for LC-MS/MS-based chiral metabolomics fingerprinting and chiral metabolomics extraction using a pair of enantiomers of chiral derivatization reagents.
Analytica chimica acta (2015-11-04)
Takahiro Takayama, Toshiki Mochizuki, Kenichiro Todoroki, Jun Zhe Min, Hajime Mizuno, Koichi Inoue, Hiroyasu Akatsu, Ichiro Noge, Toshimasa Toyo'oka
ABSTRACT
Chiral metabolites are found in a wide variety of living organisms and some of them are understood to be physiologically active compounds and biomarkers. However, the overall analysis of chiral metabolomics is quite difficult due to the high number of metabolites, the significant diversity in their physicochemical properties, and concentration range from metabolite-to-metabolite. To solve this difficulty, we developed a novel approach for chiral metabolomics fingerprinting and chiral metabolomics extraction, which is based on the labeling of a pair of enantiomers of chiral derivatization reagents (i.e., DMT-(S,R)-Pro-OSu and DMT-3(S,R)-Apy) and precursor ion scan chromatography of the derivatives. The multivariate statistics is also required for this strategy. The proposed procedures were evaluated by the detection of a diagnostic marker (i.e., d-lactic acid) using the saliva of diabetic patients. This method was used for the determination of biomarker candidates of chiral amines and carboxyls in Alzheimer's disease (AD) brain homogenates. As the results, l-phenylalanine (L-Phe) and l-lactic acid (L-LA) were identified as the decreased and increased biomarker candidates in the AD brain, respectively. Therefore, the proposed approach seems to be helpful for the determination of non-target chiral metabolomics possessing amines and carboxyls.
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Methanol solution, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
N-Acetyl-DL-tryptophan
Sigma-Aldrich
Succinic acid, ReagentPlus®, BioRenewable, ≥99.0%
Sigma-Aldrich
Succinic acid, BioReagent, BioRenewable, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Succinic acid, puriss. p.a., ACS reagent, ≥99.5% (T)
Sigma-Aldrich
4-(Dimethylamino)pyridine, ReagentPlus®, ≥99%
Sigma-Aldrich
N-Hydroxysuccinimide, 98%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
1,2-Dichloroethane, anhydrous, 99.8%
Sigma-Aldrich
Fumaric acid, FCC, FG
Sigma-Aldrich
Succinic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
DL-Valine, ≥97%
Sigma-Aldrich
1,3-Diaminopropane, ≥99%
Sigma-Aldrich
DL-Valine, ReagentPlus®, ≥99.0% (NT)
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
3-(Boc-amino)pyrrolidine, 95%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Succinic acid, BioXtra, BioRenewable, ≥99.0%
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitrile, ≥99.8% (GC)
Sigma-Aldrich
Trifluoroacetic acid, puriss. p.a., suitable for HPLC, ≥99.0% (GC)
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Trifluoroacetic acid, ReagentPlus®, 99%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Acetonitrile, ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%