Merck
  • An analytical pipeline to compare and characterise the anthocyanin antioxidant activities of purple sweet potato cultivars.
An analytical pipeline to compare and characterise the anthocyanin antioxidant activities of purple sweet potato cultivars.
Food chemistry (2015-10-17)
Yijie Hu, Liqing Deng, Jinwu Chen, Siyu Zhou, Shuang Liu, Yufan Fu, Chunxian Yang, Zhihua Liao, Min Chen
ABSTRACT
Purple sweet potato (Ipomoea batatas L.) is rich in anthocyanin pigments, which are valuable constituents of the human diet. Techniques to identify and quantify anthocyanins and their antioxidant potential are desirable for cultivar selection and breeding. In this study, we performed a quantitative and qualitative chemical analysis of 30 purple sweet potato (PSP) cultivars, using various assays to measure reducing power radical-scavenging activities, and linoleic acid autoxidation inhibition activity. Grey relational analysis (GRA) was applied to establish relationships between the antioxidant activities and the chemical fingerprints, in order to identify key bioactive compounds. The results indicated that four peonidin-based anthocyanins and three cyanidin-based anthocyanins make significant contributions to antioxidant activity. We conclude that the analytical pipeline described here represents an effective method to evaluate the antioxidant potential of, and the contributing compounds present in, PSP cultivars. This approach may be used to guide future breeding strategies.
Sigma-Aldrich
1,1-Diphenyl-2-picrylhydrazine, 97%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Trifluoroacetic acid, ≥99%, for protein sequencing
Sigma-Aldrich
Trifluoroacetic acid, ReagentPlus®, 99%
Sigma-Aldrich
Trifluoroacetic acid, puriss. p.a., suitable for HPLC, ≥99.0% (GC)
Sigma-Aldrich
2,2-Diphenyl-1-picrylhydrazyl
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, for preparative HPLC, ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%