Merck
  • Applications of Trametes versicolor crude culture filtrates in detoxification of biomass pretreatment hydrolyzates.
Applications of Trametes versicolor crude culture filtrates in detoxification of biomass pretreatment hydrolyzates.
Bioresource technology (2015-04-16)
Rajeev Kumar Kapoor, Kalavathy Rajan, Danielle Julie Carrier
ABSTRACT
Laccases have wide range of substrate specificity and find applications from pulp industry to waste water remediation. Laccases have also been used in combined pretreatment of biomass hydrolyzates to remove enzymatic and fermentation inhibitors. In this study, laccase production by Trametes versicolor strains isolated from different regions of the United States was induced using copper salts. T. versicolor crude culture filtrates (CCF), without any purification step, were tested for removal of model inhibitor compounds as well as in poplar and rice straw pretreatment hydrolyzates. Phenolic inhibitors were removed by 76% and 94% from the dilute acid hydrolyzates of rice straw and poplar, respectively, when incubated with the CCF for 12h, at room temperature. Xylo-oligosaccharide concentrations present in rice straw hydrolyzates were reduced by 64% when incubated with T. versicolor CCF. T. versicolor CCF could be a low cost technology for decreasing enzymatic and fermentation inhibitors.
Sigma-Aldrich
1-Hydroxybenzotriazole hydrate, ≥97.0% dry basis (T)
Sigma-Aldrich
Trizma® base, ≥99.0% (T)
Sigma-Aldrich
Trizma® base, BioUltra, for molecular biology, ≥99.8% (T)
Sigma-Aldrich
Salicylic acid, meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Vanillin, natural, ≥97%, FCC, FG
Sigma-Aldrich
5-(Hydroxymethyl)furfural, ≥99%, FG
Sigma-Aldrich
Salicylic acid, puriss. p.a., ≥99.0% (T)
Sigma-Aldrich
Salicylic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
5-Hydroxymethyl-2-furaldehyde, 99%
Sigma-Aldrich
Trizma® base, puriss. p.a., ≥99.7% (T)
Sigma-Aldrich
Copper(II) sulfate, anhydrous, powder, ≥99.99% trace metals basis
Sigma-Aldrich
Vanillin, ReagentPlus®, 99%
Sigma-Aldrich
Vanillic acid, 97%
Sigma-Aldrich
Salicylic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Trizma® base, BioXtra, pH 10.5-12.0 (1 M in H2O), ≥99.9% (titration)
Sigma-Aldrich
Trizma® base, BioPerformance Certified, meets EP, USP testing specifications, suitable for cell culture, ≥99.9% (titration)
Sigma-Aldrich
Salicylic acid, BioXtra, ≥99.0%
Sigma-Aldrich
Tromethamine, meets USP testing specifications
Sigma-Aldrich
Trizma® base, ≥99.9% (titration), crystalline
Sigma-Aldrich
Guaiacol, oxidation indicator
Sigma-Aldrich
Trizma® base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Sigma-Aldrich
Salicylic acid, suitable for plant cell culture
Sigma-Aldrich
p-Coumaric acid, ≥98.0% (HPLC)
Sigma-Aldrich
Salicylic acid, ≥99%, FG
Sigma-Aldrich
1-Hydroxybenzotriazole hydrate, wetted with not less than 20 wt. % water, 97% dry basis
Sigma-Aldrich
1-Hydroxybenzotriazole hydrate, wetted with not less than 14 wt. % water, 98% dry basis
Sigma-Aldrich
Vanillic acid, ≥97%, FG
Sigma-Aldrich
Guaiacol, natural, ≥99%, FG
Sigma-Aldrich
Tris(hydroxymethyl)aminomethane, ACS reagent, ≥99.8%
Sigma-Aldrich
Vanillin, ≥97%, FCC, FG