Merck
  • Runx1 is associated with breast cancer progression in MMTV-PyMT transgenic mice and its depletion in vitro inhibits migration and invasion.
Runx1 is associated with breast cancer progression in MMTV-PyMT transgenic mice and its depletion in vitro inhibits migration and invasion.
Journal of cellular physiology (2015-03-25)
Gillian Browne, Hanna Taipaleenmäki, Nicole M Bishop, Sharath C Madasu, Leslie M Shaw, Andre J van Wijnen, Janet L Stein, Gary S Stein, Jane B Lian
ABSTRACT
Runx1 is a transcription factor essential for definitive hematopoiesis, and genetic abnormalities in Runx1 cause leukemia. Runx1 is functionally promiscuous and acts as either an oncogene or tumor suppressor gene in certain epithelial cancers. Recent evidence suggests that Runx1 is an important factor in breast cancer, however, its role remains ambiguous. Here, we addressed whether Runx1 has a specific pathological role during breast cancer progression and show that Runx1 has an oncogenic function. We observed elevated Runx1 expression in a subset of human breast cancers. Furthermore, throughout the course of disease progression in a classical mouse model of breast cancer (i.e., the MMTV-PyMT transgenic model), Runx1 expression increases in the primary site (mammary gland) and is further upregulated in tumors and distal lung metastatic lesions. Ex vivo studies using tumor epithelial cells derived from these mice express significantly higher levels of Runx1 than normal mammary epithelial cells. The tumor cells exhibit increased rates of migration and invasion, indicative of an aggressive cancer phenotype. Inhibition of Runx1 expression using RNA interference significantly abrogates these cancer-relevant phenotypic characteristics. Importantly, our data establish that Runx1 contributes to murine mammary tumor development and malignancy and potentially represents a key disease-promoting and prognostic factor in human breast cancer progression and metastasis.
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Runx1
Sigma-Aldrich
Acrylamide solution, 40%, suitable for electrophoresis, sterile-filtered
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99% (HPLC), powder
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Acrylamide, for molecular biology, ≥99% (HPLC)
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
MISSION® esiRNA, targeting human RUNX1
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)