Merck
  • In-situ preparation, characterization and anticorrosion property of polypropylene glycol/silver nanoparticles composite for mild steel corrosion in acid solution.
In-situ preparation, characterization and anticorrosion property of polypropylene glycol/silver nanoparticles composite for mild steel corrosion in acid solution.
Journal of colloid and interface science (2015-10-05)
Moses M Solomon, Saviour A Umoren
ABSTRACT
A novel polypropylene glycol/silver nanoparticles (PPG/AgNPs) composite was prepared in-situ using natural honey as the reducing and capping agent. Characterization of the composite was done by UV-Vis spectroscopy, FTIR, TEM, XRD, and EDS. The TEM results reveal that the nanoparticles are spherical in shape. XRD and EDS results confirm the presence of elemental silver in the polymer matrix. The influence of the prepared composite on the corrosion inhibition of mild steel in 0.5M H2SO4 solution was studied by weight loss, electrochemical, SEM, EDS, and water contact angle measurements. Results show that PPG/AgNPs is effective inhibitor for mild steel in 0.5M H2SO4 solution and adsorbs onto the metal surface via chemisorption mechanism. Maximum inhibition efficiency of 94% is afforded by the highest studied concentration of PPG/AgNPs at 333K from weight loss measurements. Potentiodynamic polarization results reveal that the composite acts as a mixed-type corrosion inhibitor. Adsorption of PPG/AgNPs composite onto the mild steel surface follows Temkin adsorption isotherm. The SEM, EDS, and water contact angle images confirm the formation of PPG/AgNPs protective film on the mild steel surface.
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
Silver nitrate, 99.9999% trace metals basis
Sigma-Aldrich
Silver nitrate, BioXtra, >99% (titration)
Sigma-Aldrich
Silver nitrate, BioReagent, suitable for plant cell culture, >99% (titration)
Sigma-Aldrich
Silver nitrate, anhydrous, ≥99.999% trace metals basis
Sigma-Aldrich
Silver nitrate, meets analytical specification of Ph. Eur., BP, USP, 99.8-100.5%
Sigma-Aldrich
Silver nitrate, ReagentPlus®, ≥99.0% (titration)
Sigma-Aldrich
Sulfuric acid, ACS reagent, 95.0-98.0%
Sigma-Aldrich
Sulfuric acid, puriss. p.a., for determination of Hg, ACS reagent, reag. ISO, reag. Ph. Eur., 95.0-97.0%
Sigma-Aldrich
Silver nitrate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Silver nitrate, puriss. p.a., ≥99.5% (AT)
Sigma-Aldrich
Silver nitrate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sulfuric acid solution, puriss. p.a., ≥25% (T)
Sigma-Aldrich
Silver nitrate solution, 2.5 % (w/v) AgNO3 in H2O
Sigma-Aldrich
Silver nitrate on silica gel, extent of labeling: ~10 wt. % loading, +230 mesh