Merck
  • Heterocyclic amine-modified polyethylenimine as gene carriers for transfection of mammalian cells.
Heterocyclic amine-modified polyethylenimine as gene carriers for transfection of mammalian cells.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V (2015-07-26)
Zahra Salmasi, Wayne Thomas Shier, Maryam Hashemi, Elahe Mahdipour, Hamideh Parhiz, Khalil Abnous, Mohammad Ramezani
ABSTRACT
Branched polyethylenimine (PEI) is extensively used as a polycationic non-viral vector for gene delivery. Polyplexes formed with PEI are believed to be released from endocytotic vesicles by the osmotic burst mechanism in the rate-limiting step in transfection. Increasing the buffering capacity of PEI derivatives in the endosomal pH range of 4.5-7.5 should enhance transfection efficiency. In this study, PEI was derivatized by covalently attaching heterocyclic amine moieties (piperazine, pyridine and imidazole rings with pKa values from 5.23 to 6.04) through amide bonds. PEI derivatives with 50% of the primary amines on PEI exhibited increased buffering capacity, increased transfection activity and decreased cytotoxicity in murine neuroblastoma (Neuro-2a) cells. The relative effectiveness in enhancing transfection efficiency was piperazine>pyridine>histidine, but each type of amine was the most effective under a particular set of conditions. Modified vectors could significantly improve transfection efficiency in murine mesenchymal stem cells. PEI25 derivatized at 50% with histidine administered as polyplexes in the tail veins of mice resulted in remarkably enhanced luciferase gene expression in the expected organ distribution and much lower toxicity than underivatized PEI25.
Sigma-Aldrich
Dimethyl sulfoxide
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Piperazine-2-carboxylic acid dihydrochloride, 98%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Ethanesulfonic acid, 95%
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
1,2-Dichloroethane, anhydrous, 99.8%
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
3-Pyridylacetic acid hydrochloride, 98%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
1,2-Dichloroethane, ACS reagent, ≥99.0%
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
HEPES, BioUltra, ≥99.5% (T)
SAFC
L-Histidine
Sigma-Aldrich
L-Histidine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
L-Histidine, suitable for cell culture, meets EP, USP testing specifications, from non-animal source