Merck
  • Purification and characterization of lipopolysaccharides from six strains of non-O157 Shiga toxin-producing Escherichia coli.
Purification and characterization of lipopolysaccharides from six strains of non-O157 Shiga toxin-producing Escherichia coli.
Journal of microbiological methods (2015-06-21)
Loreen R Stromberg, Zachary R Stromberg, Afsheen Banisadr, Steven W Graves, Rodney A Moxley, Harshini Mukundan
ABSTRACT
Certain Shiga toxin-producing Escherichia coli (STEC) are virulent human pathogens that are most often acquired through contaminated food. The United States Department of Agriculture, Food Safety and Inspection Service has declared several serogroups of STEC as adulterants in non-intact raw beef products. Hence, sensitive and specific tests for the detection of these STEC are a necessity for implementation in food safety programs. E. coli serogroups are identified by their respective O-antigen moiety on the lipopolysaccharide (LPS) macromolecule. We propose that the development of O-antigen-specific immunological assays can facilitate simple and rapid discriminatory detection of STEC in beef. However, the resources (antigens and antibodies) required for such development are not readily available. To overcome this, we extracted and characterized LPS and O-antigen from six STEC strains. Using hot phenol extraction, we isolated the LPS component from each strain and purified it using a series of steps to eliminate proteins, nucleic acids, and lipid A antigens. Antigens and crude LPS extracts were characterized using gel electrophoresis, immunoblotting, and modified Western blotting with commercially available antibodies, thus assessing the serogroup specificity and sensitivity of available ligands as well. The results indicate that, while many commercially available antibodies bind LPS, their activities and specificities are highly variable, and often not as specific as those required for serogroup discrimination. This variability could be minimized by the production of antibodies specific for the O-antigen. Additionally, the antigens generated from this study provide a source of characterized LPS and O-antigen standards for six serogroups of STEC.
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
SAFC
Glycine
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
Adenosine 5′-phosphosulfate sodium salt, ≥85%
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Acrylamide, for molecular biology, ≥99% (HPLC)
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99% (HPLC), powder
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Glycine, BioUltra, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Eosin B, certified by the Biological Stain Commission, Dye content 90 %
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Sigma-Aldrich
Phenol, contains hypophosphorous as stabilizer, loose crystals, ACS reagent, ≥99.0%
Sigma-Aldrich
Phenol, puriss., ≥99.5% (GC), meets analytical specification of Ph. Eur., BP, USP, crystalline (detached)
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Sigma-Aldrich
Eosin B, Dye content 95 %
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioUltra, for molecular biology, ≥99.0% (AT)
Sigma-Aldrich
Phenol, ≥96.0% (calc. on dry substance, T)
Sigma-Aldrich
Ammonium persulfate, BioUltra, for molecular biology, ≥98.0% (RT)
Sigma-Aldrich
Phenol, ≥99%
Sigma-Aldrich
Phenol, BioUltra, for molecular biology, TE-saturated, ~73% (T)
Sigma-Aldrich
Phenol solution, BioReagent, Saturated with 0.01 M citrate buffer, pH 4.3 ± 0.2, for molecular biology