Merck
  • Effect of the oxidation treatment on the production of cellulose nanofiber suspensions from Posidonia oceanica: The rheological aspect.
Effect of the oxidation treatment on the production of cellulose nanofiber suspensions from Posidonia oceanica: The rheological aspect.
Carbohydrate polymers (2015-10-03)
Fedia Bettaieb, Oleksandr Nechyporchuk, Ramzi Khiari, Mohamed Farouk Mhenni, Alain Dufresne, Mohamed Naceur Belgacem
ABSTRACT
Different grades of cellulose nanofibrils (CNF) were prepared from Posidonia oceanica balls and leaves (POB and POL). Pretreatment using 2,2,6,6-tetramethylpiperidine-1-oxyl (TEMPO)-mediated oxidation was performed to facilitate the fibrillation during ultrafine friction grinding process. The ensuing CNF batches were compared in terms of morphology and degree of fibrillation. The rheological properties of the produced CNF suspensions were also analyzed for varying doses of sodium hypochlorite used during the TEMPO- mediated oxidation procedure. The stronger fibrous network structures were formed when increasing the oxidant concentration, which was confirmed by the increase of the storage moduli value. P. oceanica balls were found to undergo stronger fibrillation and, consequently, to form stronger networks, compared to P. oceanica leaves, when using equivalent concentration of the oxidizing agent.
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Sodium hypochlorite solution, purum, ~10% (RT)
Sigma-Aldrich
Acetic acid, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.8-100.5%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid, glacial, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid, glacial, puriss., 99-100%
Sigma-Aldrich
Sodium hypochlorite solution, reagent grade, available chlorine 10-15 %
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Sodium hypochlorite solution, reagent grade, available chlorine 4.00-4.99 %
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium chlorite, puriss. p.a., 80% (RT)
Sigma-Aldrich
Sodium chlorite, technical grade, 80%