Merck
  • Antimicrobial Efficacy of a Lactic Acid and Citric Acid Blend against Shiga Toxin-Producing Escherichia coli, Salmonella, and Nonpathogenic Escherichia coli Biotype I on Inoculated Prerigor Beef Carcass Surface Tissue.
Antimicrobial Efficacy of a Lactic Acid and Citric Acid Blend against Shiga Toxin-Producing Escherichia coli, Salmonella, and Nonpathogenic Escherichia coli Biotype I on Inoculated Prerigor Beef Carcass Surface Tissue.
Journal of food protection (2015-11-29)
Brittney R Scott, Xiang Yang, Ifigenia Geornaras, Robert J Delmore, Dale R Woerner, Jeremy M Adler, Keith E Belk
ABSTRACT
Studies were conducted to (i) determine whether inoculants of nonpathogenic Escherichia coli biotype I effectively served as surrogates for E. coli O157:H7, non-O157 Shiga toxin-producing E. coli, and Salmonella when prerigor beef carcass tissue was treated with a commercially available blend of lactic acid and citric acid (LCA) at a range of industry conditions of concentration, temperature, and pressure; (ii) determine the antimicrobial efficacy of LCA; and (iii) investigate the use of surrogates to validate a hot water and LCA sequential treatment as a carcass spray intervention in a commercial beef harvest plant. In an initial laboratory study, beef brisket tissue samples were left uninoculated or were inoculated (∼6 log CFU/cm(2)) on the adipose side with E. coli O157:H7 (5-strain mixture), non-O157 Shiga toxin-producing E. coli (12-strain mixture), Salmonella (6-strain mixture), or nonpathogenic E. coli (5-strain mixture). Samples were left untreated (control) or were treated with LCA, in a spray cabinet, at one of eight combinations of solution concentration (1.9 and 2.5%), solution temperature (43 and 60°C), and application pressure (15 and 30 lb/in(2)). In a second study, the E. coli surrogates were inoculated (∼6 log CFU/cm(2)) on beef carcasses in a commercial facility to validate the use of a hot water treatment (92.2 to 92.8°C, 13 to 15 lb/in(2)) followed by an LCA treatment (1.9%, 50 to 51.7°C, 13 to 15 lb/in(2), 10 s). In the in vitro study, surrogate and pathogen bacteria did not differ in their response to the tested LCA treatments. Treatment with LCA reduced (P < 0.05) inoculated populations by 0.9 to 1.5 log CFU/cm(2), irrespective of inoculum type. The hot water and LCA sequential treatments evaluated in the commercial facility reduced (P < 0.05) the inoculated nonpathogenic E. coli surrogates on carcasses by 3.7 log CFU/cm(2). This study therefore provides the meat industry with data for this sequential multiple hurdle system for the operation parameters described.
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Glycerol solution, puriss., meets analytical specification of Ph. Eur., BP, 84-88%
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
Glycerol, BioUltra, for molecular biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glycerol, puriss., anhydrous, 99.0-101.0% (alkalimetric)
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerol, puriss. p.a., ACS reagent, anhydrous, dist., ≥99.5% (GC)