Merck
  • Characterization and immunomodulatory activities of polysaccharides from Spirogyra neglecta (Hassall) Kützing.
Characterization and immunomodulatory activities of polysaccharides from Spirogyra neglecta (Hassall) Kützing.
Bioscience, biotechnology, and biochemistry (2015-05-15)
Utoomporn Surayot, JianGuo Wang, Ju Hun Lee, Chartchai Kanongnuch, Yuwadee Peerapornpisal, SangGuan You
ABSTRACT
Sulfated polysaccharides (SP) isolated from freshwater green algae, Spirogyra neglecta (Hassall) Kützing, and fractionated SPs were examined to investigate their molecular characteristics and immunomodulatory activity. The crude and fractionated SPs (F1, F2, and F3) consisted mostly of carbohydrates (68.5-85.3%), uronic acids (3.2-4.9%), and sulfates (2.2-12.2%) with various amounts of proteins (2.6-17.1%). D-galactose (23.5-27.3%), D-glucose (11.5-24.8%), L-fucose (19.0-26.7%), and L-rhamnose (16.4-18.3%) were the major monosaccharide units of these SPs with different levels of L-arabinose (3.0-9.4%), D-xylose (4.6-9.8%), and D-mannose (0.4-2.3%). The SPs contained two sub-fractions with molecular weights (Mw) ranging from 164 × 10(3) to 1460 × 10(3) g/mol. The crude and fractionated SPs strongly stimulated murine macrophages, producing considerable amounts of nitric oxide and various cytokines via up-regulation of their mRNA expression by activation of nuclear factor-kappa B and mitogen-activated protein kinases pathways. The main backbone of the most immunoenhancing SP was (1→3)-L-Fucopyranoside, (1→4,6)-D-Glucopyranoside, and (1→4)-D-Galactopyranoside.
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Sigma-Aldrich
Sodium borodeuteride, 98 atom % D, 90% (CP)
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Acetic anhydride, Arxada quality, ≥99.5% (GC)
Sigma-Aldrich
Acetic anhydride, 99.5%
Sigma-Aldrich
Acetic anhydride, ACS reagent, ≥98.0%
Sigma-Aldrich
Acetic anhydride, ReagentPlus®, ≥99%
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
Glycerol, BioUltra, for molecular biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glycerin, meets USP testing specifications