Merck
  • Experimental investigation on the soil sorption properties and hydrophobicity of polymethoxylated, polyhydroxylated diphenyl ethers and methoxylated-, hydroxylated-polychlorinated diphenyl ethers.
Experimental investigation on the soil sorption properties and hydrophobicity of polymethoxylated, polyhydroxylated diphenyl ethers and methoxylated-, hydroxylated-polychlorinated diphenyl ethers.
Chemosphere (2015-04-29)
Xuesheng Zhang, Xiaolan Zeng, Li Qin, Ruijuan Qu, Jiaqi Shi, Zhongbo Wei, Shaogui Yang, Zunyao Wang
ABSTRACT
In the present study, twenty-six types of polymethoxylated diphenyl ethers (PMeODEs), twenty types of polyhydroxylated diphenyl ethers (PHODEs), seven types of methoxylated-polychlorinated diphenyl ethers (MeO-PCDEs) and seven types of hydroxylated-polychlorinated diphenyl ethers (HO-PCDEs) were synthesized. The logKow and logKoc values of all of the synthesized compounds were then determined using HPLC. The soil sorption properties of five types of selected substituted diphenyl ethers (DEs) were investigated. Sorption behavior studies suggested that rapid sorption played a primary role in the sorption process of the selected DEs and their sorption isotherms were fitted the Freundlich logarithmic model. For PMeODEs and PHODEs, with the increase in the number of substituents, both logKow and logKoc values exhibited linearly decreasing trends. Unlike PMeODEs and PHODEs, both logKow and logKoc values of MeO/HO-PCDEs were decreased linearly with the increasing number of chlorine atoms. The reason maybe that both methoxy and hydroxyl are hydrophilic groups, whereas the chlorine atom is hydrophobic group. Linear relationships were observed for the logKow and logKoc of all studied DEs. Moreover, the logKow of PMeODEs, PHODEs, MeO- and HO-PCDEs and their corresponding PCDEs showed good linearity.
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D, contains 1 % (v/v) TMS
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Dimethyl sulfoxide-d6, anhydrous, 99.9 atom % D
Sigma-Aldrich
Cesium carbonate, ReagentPlus®, 99%
Sigma-Aldrich
Cesium carbonate, 99.995% trace metals basis
Sigma-Aldrich
Copper(I) iodide, purum, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.5 atom % D
Sigma-Aldrich
Cesium carbonate, 99.9% trace metals basis
Sigma-Aldrich
1,4-Diaminobutane, 99%
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Copper(I) iodide, 99.999% trace metals basis
Sigma-Aldrich
Cesium carbonate, 99.95% trace metals basis
Sigma-Aldrich
Dimethyl sulfoxide-d6, "Special HOH", ≥99.9 atom % D
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Copper(I) iodide, anhydrous, 99.995% trace metals basis
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Magnesium sulfate solution, BioUltra, for molecular biology
Sigma-Aldrich
Magnesium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%
Sigma-Aldrich
Hexane, anhydrous, 95%
Sigma-Aldrich
Magnesium sulfate, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Magnesium sulfate solution, for molecular biology, 1.00 M±0.04 M
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.