Merck
  • Different Responses in MMP/TIMP Expression of U937 and HepG2 Cells to Dengue Virus Infection.
Different Responses in MMP/TIMP Expression of U937 and HepG2 Cells to Dengue Virus Infection.
Japanese journal of infectious diseases (2015-02-13)
Pannatas Seanpong, Chanya Srisaowakarn, Anothai Thammaporn, Vijittra Leardkamolkarn, Supeecha Kumkate
ABSTRACT
Disease severities following dengue virus (DV) infection are the result of increased vascular permeability leading to hypovolemic shock. Matrix metalloproteinases (MMPs) are believed to play a key role in promoting such severities. A previous study reported that supernatants of DV-infected dendritic cells (DCs), which contained high levels of MMP-2 and MMP-9, induced vascular leakage in a mouse model. In the present study, we investigated whether hepatocytes (HepG2) and monocytes (U937) could be additional sources of MMPs during DV infection. HepG2 and U937 cells were exposed to DV serotype 2 strain 16681. The secretion of MMP-2 and MMP-9 was detected using gelatin zymography. We found that DV infection in the HepG2 cells promoted MMP-2 production while that in the U937 cells promoted MMP-9 production. Semi-quantitative RT-PCR results also confirmed that DV infection in the HepG2 cells up-regulated the expression of MMP-2 mRNA, whereas that in the U937 cells enhanced the expression of MMP-9 mRNA. We monitored the expression of endogenous TIMP-1 and TIMP-2. DV infection induced TIMP-1 expression in the U937 cells. However, lower expression of TIMP-2 was observed in the infected HepG2 cells. We believed that following DV infection, monocytes and hepatocytes can act as MMP-9 and MMP-2 producers, respectively. Their responses could be attributed to the disturbance of TIMP expression by DV in different cell types.
Product Description
SAFC
Sodium chloride solution, 5 M
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Anti-TIMP-1 Antibody, clone 102D1, clone 102D1, Chemicon®, from mouse
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis