Merck
  • Polycyclic aromatic hydrocarbons in the dagang oilfield (china): distribution, sources, and risk assessment.
Polycyclic aromatic hydrocarbons in the dagang oilfield (china): distribution, sources, and risk assessment.
International journal of environmental research and public health (2015-05-29)
Haihua Jiao, Xiaoping Rui, Shanghua Wu, Zhihui Bai, Xuliang Zhuang, Zhanbin Huang
ABSTRACT
The levels of 16 polycyclic aromatic hydrocarbons (PAHs) were investigated in 27 upper layer (0-25 cm) soil samples collected from the Dagang Oilfield (China) in April 2013 to estimate their distribution, possible sources, and potential risks posed. The total concentrations of PAHs (∑PAHs) varied between 103.6 µg·kg(-1) and 5872 µg·kg(-1), with a mean concentration of 919.8 µg·kg(-1); increased concentrations were noted along a gradient from arable desert soil (mean 343.5 µg·kg(-1)), to oil well areas (mean of 627.3 µg·kg(-1)), to urban and residential zones (mean of 1856 µg·kg(-1)). Diagnostic ratios showed diverse source of PAHs, including petroleum, liquid fossil fuels, and biomass combustion sources. Combustion sources were most significant for PAHs in arable desert soils and residential zones, while petroleum sources were a significant source of PAHs in oilfield areas. Based ontheir carcinogenity, PAHs were classified as carcinogenic (B) or not classified/non-carcinogenic (NB). The total concentrations of carcinogenic PAHs (∑BPAHs) varied from 13.3 µg·kg(-1) to 4397 µg·kg(-1) across all samples, with a mean concentration of 594.4 µg·kg(-1). The results suggest that oilfield soil is subject to a certain level of ecological environment risk.
Sigma-Aldrich
Perylene, sublimed grade, ≥99.5%
Sigma-Aldrich
Perylene, ≥99%
Sigma-Aldrich
Anthracene, sublimed grade, ≥99%
Sigma-Aldrich
Pyrene, puriss. p.a., for fluorescence, ≥99.0% (GC)
Sigma-Aldrich
Anthracene, reagent grade, 97%
Sigma-Aldrich
Anthracene, suitable for scintillation, ≥99.0% (GC)
Sigma-Aldrich
Perylene-d12, 98 atom % D
Sigma-Aldrich
Pyrene, 98%
Sigma-Aldrich
Anthracene, ReagentPlus®, 99%
Sigma-Aldrich
Pyrene, sublimed grade, 99%