Merck
  • Type-2 diabetes increases autophagy in the human heart through promotion of Beclin-1 mediated pathway.
Type-2 diabetes increases autophagy in the human heart through promotion of Beclin-1 mediated pathway.
International journal of cardiology (2015-09-20)
Pujika Emani Munasinghe, Federica Riu, Parul Dixit, Midori Edamatsu, Pankaj Saxena, Nathan S J Hamer, Ivor F Galvin, Richard W Bunton, Sharon Lequeux, Greg Jones, Regis R Lamberts, Costanza Emanueli, Paolo Madeddu, Rajesh Katare
ABSTRACT
Diabetes promotes progressive loss of cardiac cells, which are replaced by a fibrotic matrix, resulting in the loss of cardiac function. In the current study we sought to identify if excessive autophagy plays a major role in inducing this progressive loss. Immunofluorescence and western blotting analysis of the right atrial appendages collected from diabetic and non-diabetic patients undergoing coronary artery bypass graft surgery showed a marked increase in the level of autophagy in the diabetic heart, as evidenced by increased expression of autophagy marker LC3B-II and its mediator Beclin-1 and decreased expression of p62, which incorporates into autophagosomes to be efficiently degraded. Moreover, a marked activation of pro-apoptotic caspase-3 was observed. Electron microscopy showed increased autophagosomes in the diabetic heart. In vivo measurement of autophagic flux by choloroquine injection resulted in further enhancement of LC3B-II in the diabetic myocardium, confirming increased autophagic activity in the type-2 diabetic heart. Importantly, in-vitro genetic depletion of beclin-1 in high glucose treated adult rat cardiomyocytes markedly inhibited the level of autophagy and subsequent apoptotic cell death. These findings demonstrate the pathological role of autophagy in the type-2 diabetic heart, opening up a potentially novel therapeutic avenue for the treatment of diabetic heart disease.
Sigma-Aldrich
MISSIONĀ® esiRNA, targeting human BECN1
Sigma-Aldrich
Glutaric dialdehyde solution, 50 wt. % in H2O, FCC
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
Glutaraldehyde solution, Grade II, 25% in H2O
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
MISSIONĀ® esiRNA, targeting mouse Becn1
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, 50% in H2O, suitable for photographic applications
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O