Merck
  • Selective Activation of C=C Bond in Sustainable Phenolic Compounds from Lignin via Photooxidation: Experiment and Density Functional Theory Calculations.
Selective Activation of C=C Bond in Sustainable Phenolic Compounds from Lignin via Photooxidation: Experiment and Density Functional Theory Calculations.
Photochemistry and photobiology (2015-08-14)
Morgan Zielinski Goldberg, Luke A Burke, Alexander Samokhvalov
ABSTRACT
Lignocellulosic biomass can be converted to high-value phenolic compounds, such as food additives, antioxidants, fragrances and fine chemicals. We investigated photochemical and heterogeneous photocatalytic oxidation of two isomeric phenolic compounds from lignin, isoeugenol and eugenol, in several nonprotic solvents, for the first time by experiment and the density functional theory (DFT) calculations. Photooxidation was conducted under ambient conditions using air, near-UV light and commercial P25 TiO2 photocatalyst, and the products were determined by TLC, UV-Vis absorption spectroscopy, HPLC-UV and HPLC-MS. Photochemical and photocatalytic oxidation of isoeugenol proceeds via the mild oxidative "dimerization" to produce the lignan dehydrodiisoeugenol (DHDIE), while photooxidation of eugenol does not proceed. The DFT calculations suggest a radical stepwise mechanism for the oxidative "dimerization" of isoeugenol to DHDIE as was calculated for the first time.
Sigma-Aldrich
Vanillin, natural, ≥97%, FCC, FG
Sigma-Aldrich
Eugenol, ≥98%, FCC, FG
Sigma-Aldrich
Titanium(IV) oxide, rutile, 99.995% trace metals basis
Sigma-Aldrich
N,N-Dimethylformamide, anhydrous, 99.8%
Sigma-Aldrich
Eugenol, ReagentPlus®, 99%
Sigma-Aldrich
Isoeugenol, mixture of cis and trans, 99%, FG
Sigma-Aldrich
Vanillin, ReagentPlus®, 99%
Sigma-Aldrich
Isoeugenol, 98%, mixture of cis and trans
Sigma-Aldrich
Titanium(IV) oxide, rutile, ≥99.98% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, rutile, powder, <5 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, rutile, nanopowder, <100 nm particle size, 99.5% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, rutile, <001>, (single crystal substrate), ≥99.9% trace metals basis, L × W × thickness 10 mm × 10 mm × 0.5 mm
Sigma-Aldrich
Titanium(IV) oxide, mixture of rutile and anatase, nanopowder, <100 nm particle size (BET), 99.5% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, mixture of rutile and anatase, nanoparticles, <150 nm particle size (volume distribution, DLS), dispersion, 40 wt. % in H2O, 99.5% trace metals basis
Sigma-Aldrich
Isoeugenol, natural, 99%, FG
Sigma-Aldrich
N,N-Dimethylformamide, for molecular biology, ≥99%
Sigma-Aldrich
Eugenol, natural, ≥98%, FG
Sigma-Aldrich
Vanillin, ≥97%, FCC, FG
Sigma-Aldrich
N,N-Dimethylformamide, biotech. grade, ≥99.9%
Sigma-Aldrich
N,N-Dimethylformamide, ACS reagent, ≥99.8%
Sigma-Aldrich
N,N-Dimethylformamide, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
N,N-Dimethylformamide, ReagentPlus®, ≥99%
Sigma-Aldrich
Titanium(IV) oxide, nanopowder, 21 nm primary particle size (TEM), ≥99.5% trace metals basis
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Titanium(IV) oxide, anatase, powder, −325 mesh, ≥99% trace metals basis
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Titanium(IV) oxide, anatase, powder, 99.8% trace metals basis
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture