Merck
  • Synthesis and Nematicidal Activities of 1,2,3-Benzotriazin-4-one Derivatives against Meloidogyne incognita.
Synthesis and Nematicidal Activities of 1,2,3-Benzotriazin-4-one Derivatives against Meloidogyne incognita.
Journal of agricultural and food chemistry (2015-07-07)
Gaolei Wang, Xiulei Chen, Yayun Deng, Zhong Li, Xiaoyong Xu
ABSTRACT
A series of novel 1,2,3-benzotriazin-4-one derivatives were synthesized by the reaction of 3-bromoalkyl-1,2,3-benzotriazin-4-ones with potassium salt of 2-cyanoimino-4-oxothiazolidine in the presence of potassium iodide. Nematicidal assays in vivo showed that some of them exhibited good control efficacy against the cucumber root-knot nematode disease caused by Meloidogyne incognita, up to 100% at the concentration of 10.0 mg L(-1), which indicated that 1,2,3-benzotriazin-4-one derivatives might be potential for novel promising nematicides. The nematicidal activity was influenced by the combination of substituent type, substituted position, and linker length in the molecule. The inhibition rate data at the concentrations of 5.0 and 1.0 mg L(-1) for the compounds with high inhibitory activities were also provided. When tested in vitro, none of them showed direct inhibition against M. incognita. The investigation of a significant difference between in vivo and in vitro data is in progress.
Sigma-Aldrich
Potassium carbonate, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Potassium carbonate, meets USP testing specifications
Sigma-Aldrich
Potassium carbonate, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Sodium nitrite, 99.999% trace metals basis
Sigma-Aldrich
1,6-Dibromohexane, 96%
Sigma-Aldrich
Potassium carbonate, 99.995% trace metals basis
Sigma-Aldrich
1,4-Dibromobutane, 99%
Sigma-Aldrich
1,5-Dibromopentane, 97%
Sigma-Aldrich
Potassium hydroxide, anhydrous, ≥99.95% trace metals basis
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.5 atom % D
Sigma-Aldrich
Potassium iodide, ≥99.99% trace metals basis
Sigma-Aldrich
1,3-Dibromopropane, ReagentPlus®, 99%
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Potassium iodide, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D, contains 1 % (v/v) TMS
Sigma-Aldrich
Sodium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Potassium iodide, suitable for plant cell culture
Sigma-Aldrich
Sodium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium iodide, BioXtra, ≥99.0%
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Sodium sulfate, ≥99.0%, suitable for plant cell culture
Sigma-Aldrich
Sodium sulfate, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Dimethyl sulfoxide-d6, anhydrous, 99.9 atom % D
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)