Merck
  • FTY720 enhances TRAIL-mediated apoptosis by up-regulating DR5 and down-regulating Mcl-1 in cancer cells.
FTY720 enhances TRAIL-mediated apoptosis by up-regulating DR5 and down-regulating Mcl-1 in cancer cells.
Oncotarget (2015-04-07)
Seon Min Woo, Bo Ram Seo, Kyoung-jin Min, Taeg Kyu Kwon
ABSTRACT
FTY720, Fingolimod, is a functional antagonist to the sphingosine-1-phosphate (S1P) receptor and an inhibitor of sphingosine kinase 1. Here, we showed that a combination of FTY720 and TRAIL induced apoptosis in human renal, breast, and colon carcinoma cells. Most importantly, this combination had no effect on normal cells. Furthermore, the combined treatment with FTY720 and TRAIL reduced tumor growth in xenograft models. FTY720 up-regulated death receptor (DR)5 at post-translational level. Knockdown of DR5 markedly blocked apoptosis induced by the combined treatment. FTY720 also inhibited Mcl-1 expression at the post-translational level. Over-expression of Mcl-1 blocked apoptosis induced by FTY720 and TRAIL. Interestingly, phospho-FTY720 and inhibitors of sphingosine kinase failed to enhance TRAIL-induced apoptosis. Thus, FTY720 enables TRAIL-induced apoptosis through up-regulation of DR5 and down-regulation of Mcl-1 in human cancer cells.
Sigma-Aldrich
Glycine ethyl ester hydrochloride, 99%
Sigma-Aldrich
meso-2,3-Dimercaptosuccinic acid, ~98%
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Tnfrsf10b
Sigma-Aldrich
Lactacystin, ≥90% (HPLC)
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Mcl1
Sigma-Aldrich
MISSION® esiRNA, targeting human TNFRSF10B
Sigma-Aldrich
MISSION® esiRNA, targeting human MCL1
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Dimethyl sulfide, natural, ≥99%, FCC, FG
Sigma-Aldrich
Dimethyl sulfide, redistilled, ≥99%, FCC, FG
Sigma-Aldrich
Dimethyl sulfide, ≥99%
Sigma-Aldrich
Dimethyl sulfide, anhydrous, ≥99.0%
Sigma-Aldrich
Fluorescein, for fluorescence, free acid
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Sigma-Aldrich
D-Sphingosine, synthetic
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
N-Acetyl-L-cysteine, BioXtra, ≥99% (TLC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
N-Acetyl-L-cysteine, Sigma Grade, ≥99% (TLC), powder
Sigma-Aldrich
N-Acetyl-L-cysteine, BioReagent, suitable for cell culture
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Glycerol, BioUltra, for molecular biology, anhydrous, ≥99.5% (GC)