Merck
  • An improved odor bait for monitoring populations of Aedes aegypti-vectors of dengue and chikungunya viruses in Kenya.
An improved odor bait for monitoring populations of Aedes aegypti-vectors of dengue and chikungunya viruses in Kenya.
Parasites & vectors (2015-05-01)
Eunice A Owino, Rosemary Sang, Catherine L Sole, Christian Pirk, Charles Mbogo, Baldwyn Torto
ABSTRACT
Effective surveillance and estimation of the biting fraction of Aedes aegypti is critical for accurate determination of the extent of virus transmission during outbreaks and inter-epidemic periods of dengue and chikungunya fever. Here, we describe the development and use of synthetic human odor baits for improved sampling of adult Ae. aegypti, in two dengue and chikungunya fevers endemic areas in Kenya; Kilifi and Busia counties. We collected volatiles from the feet and trunks of two female and two male volunteers aged between 25 and 45 years. We used coupled gas chromatography- electroantennographic detection (GC/EAD) analysis to screen for antennally-active components from the volatiles and coupled GC-mass spectrometry (GC/MS) to identify the EAD-active components. Using randomized replicated designs, we compared the efficacies of Biogents (BG) sentinel traps baited with carbon dioxide plus either single or blends of the identified compounds against the BG sentinel trap baited with carbon dioxide plus the BG commercial lure in trapping Ae. aegypti. The daily mosquito counts in the different traps were subjected to negative binomial regression following the generalized linear models procedures. A total of ten major EAD-active components identified by GC/MS as mainly aldehydes and carboxylic acids, were consistently isolated from the human feet and trunk volatiles from at least two volunteers. Field assays with synthetic chemicals of the shared EAD-active components identified from the feet and trunk gave varying results. Ae. aegypti were more attracted to carbon dioxide baited BG sentinel traps combined with blends of aldehydes than to similar traps combined with blends of carboxylic acids. When we assessed the efficacy of hexanoic acid detected in odors of the BG commercial lure and volunteers plus carbon dioxide, trap captures of Ae. aegypti doubled over the trap baited with the commercial BG lure. However, dispensing aldehydes and carboxylic acids together in blends, reduced trap captures of Ae. aegypti by ~45%-50%. Our results provide evidence for roles of carboxylic acids and aldehydes in Ae. aegypti host attraction and also show that of the carboxylic acids, hexanoic acid is a more effective lure for the vector than the BG commercial lure.
Sigma-Aldrich
Nonanal, 95%
Sigma-Aldrich
Hexanal, 98%
Sigma-Aldrich
Decanal, ≥98% (GC), liquid
Sigma-Aldrich
Propionic acid, BioReagent, suitable for insect cell culture, ~98%
Sigma-Aldrich
Propionic acid, natural, 99%, FG
Sigma-Aldrich
Hexanal, ≥97%, FCC, stabilized, FG
Sigma-Aldrich
Isovaleric acid, natural, ≥98%, FG
Sigma-Aldrich
Hexanal, natural, ≥95%, FG
Sigma-Aldrich
Isovaleric acid, ≥99%, FCC, FG
Sigma-Aldrich
Nonanal, ≥95%, FCC
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Propionic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Decanal, natural, ≥97%, FG
Sigma-Aldrich
Isovaleric acid, 99%
Sigma-Aldrich
Propionic acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Decanal, FCC, FG
Sigma-Aldrich
Propionic acid, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Nonanal, natural, ≥98%, FG
Sigma-Aldrich
Octanal, natural, ≥95%, FCC, FG
Sigma-Aldrich
Hexanoic acid, ≥98%, FCC, FG
Sigma-Aldrich
Butylated hydroxytoluene, ≥99%, FCC, FG
Sigma-Aldrich
Octanal, ≥95%, FCC, FG
Sigma-Aldrich
Heptaldehyde, 95%
Sigma-Aldrich
Octanal, 99%
Sigma-Aldrich
Hexanoic acid, ≥99%
Sigma-Aldrich
Heptanal, ≥95%, FCC, FG