Merck
  • Ultrasound-Triggered Phase-Transition Cationic Nanodroplets for Enhanced Gene Delivery.
Ultrasound-Triggered Phase-Transition Cationic Nanodroplets for Enhanced Gene Delivery.
ACS applied materials & interfaces (2015-05-29)
Di Gao, Ming Xu, Zhong Cao, Jinbiao Gao, Ya Chen, Yingqin Li, Zhe Yang, Xiaoyan Xie, Qing Jiang, Wei Wang, Jie Liu
ABSTRACT
Ultrasound as an external stimulus for enhanced gene transfection represents a safe, noninvasive, cost-effective delivery strategy for gene therapy. Herein, we have developed an ultrasound-triggered phase-transition cationic nanodroplet based on a novel perfluorinated amphiphilic poly(amino acid), which could simultaneously load perfluoropentane (PFP) and nucleic acids. The heptadecafluoroundecylamine (C11F17-NH2) was chosen to initiate β-benzyl-L-aspartate N-carboxyanhydride (BLA-NCA) ring-opening polymerization to prepare C11F17-poly(β-benzyl-L-aspartate) (C11F17-PBLA). Subsequently, C11F17-poly{N-[N'-(2-aminoethyl)]aspartamide} [C11F17-PAsp(DET)] was synthesized by aminolysis reaction of C11F17-PBLA with diethylenetriamine (DET). PFP/pDNA-loaded nanodroplets PFP-TNDs [PFP/C11F17-PAsp(DET)/LucDNA/γ-PGA or poly(glutamic acid)-g-MeO-poly(ethylene glycol) (PGA-g-mPEG) ternary nanodroplets] were primarily formulated by an oil/water emulsification method, followed by surface modification with PGA-g-mPEG. The average diameter of PFP-TNDs ranged from 300 to 400 nm, and transmission electron microscopy images showed that the nanodroplets were nearly spherical in shape. The ζ potential of the nanodroplets dramatically decreased from +54.3 to +15.3 mV after modification with PGA-g-mPEG, resulting in a significant increase of the stability of the nanodroplets in the serum-containing condition. With ultrasound irradiation, the gene transfection efficiency was enhanced 14-fold on HepG2 cells, and ultrasound-triggered phase-transition cationic nanodroplets also displayed a good ultrasound contrast effect. These results suggest that the PFP/DNA-loaded phase-transition cationic nanodroplets can be utilized as efficient theranostic agents for targeting gene delivery.
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, "100%", 99.96 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D, contains 1 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 1 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, anhydrous
Sigma-Aldrich
Dimethyl sulfoxide-d6, anhydrous, 99.9 atom % D
Sigma-Aldrich
Chloroform-d, "100%", 99.95 atom % D
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.05 % (v/v) TMS
Sigma-Aldrich
Diethylenetriamine, ReagentPlus®, 99%
Sigma-Aldrich
Bis(trichloromethyl) carbonate, purum, ≥99.0% (AT)
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.5 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, contains 0.5 wt. % silver foil as stabilizer, 0.03 % (v/v) TMS
Sigma-Aldrich
1-Methyl-2-pyrrolidinone, anhydrous, 99.5%
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.1 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, 99.8 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D
Sigma-Aldrich
Chloroform-d, "100%", 99.96 atom % D, contains 0.5 wt. % silver wire as stabilizer
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, contains 0.5 wt. % silver foil as stabilizer
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Dimethyl sulfoxide-d6, "Special HOH", ≥99.9 atom % D
Sigma-Aldrich
Fluorescein, for fluorescence, free acid
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 0.75 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
Sigma-Aldrich
Triphosgene, reagent grade, 98%
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 0.05 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt