Merck
  • Cysteinyl leukotrienes mediate lymphokine killer activity induced by NKG2D and IL-15 in cytotoxic T cells during celiac disease.
Cysteinyl leukotrienes mediate lymphokine killer activity induced by NKG2D and IL-15 in cytotoxic T cells during celiac disease.
The Journal of experimental medicine (2015-08-26)
Fangming Tang, Benjamin Sally, Kathryn Lesko, Valentina Discepolo, Valerie Abadie, Cezary Ciszewski, Carol Semrad, Stefano Guandalini, Sonia S Kupfer, Bana Jabri
ABSTRACT
Eicosanoids are inflammatory mediators that play a key but incompletely understood role in linking the innate and adaptive immune systems. Here, we show that cytotoxic effector T cells (CTLs) are capable of both producing and responding to cysteinyl leukotrienes (CystLTs), allowing for the killing of target cells in a T cell receptor-independent manner. This process is dependent on the natural killer receptor NKG2D and exposure to IL-15, a cytokine induced in distressed tissues. IL-15 and NKG2D signaling drives the up-regulation of key enzymes implicated in the synthesis of CystLTs, as well as the expression of CystLT receptors, suggesting a positive feedback loop. Finally, although the CystLT pathway has been previously linked to various allergic disorders, we provide unexpected evidence for its involvement in the pathogenesis of celiac disease (CD), a T helper 1 cell-mediated enteropathy induced by gluten. These findings provide new insights into the cytolytic signaling pathway of NKG2D and the pathogenesis of organ-specific immune disorders. Furthermore, they suggest that the blockade of CystLT receptors may represent a potent therapeutic target for CD or potentially other autoimmune disorders in which NKG2D has been implicated.
Sigma-Aldrich
Hematoxylin
Sigma-Aldrich
Hematoxylin, certified by the BSC
Sigma-Aldrich
Anti-Phosphotyrosine Antibody, clone 4G10®, clone 4G10®, Upstate®, from mouse