Merck
  • Simultaneous analysis of Alternaria toxins and citrinin in tomato: an optimised method using liquid chromatography-tandem mass spectrometry.
Simultaneous analysis of Alternaria toxins and citrinin in tomato: an optimised method using liquid chromatography-tandem mass spectrometry.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment (2015-07-28)
Ádám Tölgyesi, Joerg Stroka, Vytautas Tamosiunas, Theresa Zwickel
ABSTRACT
Alternaria toxins and citrinin are mycotoxins produced by fungi growing on different raw materials and agricultural commodities. Maximum levels of these toxins in foods are currently under consideration by the European Commission as a risk management measure. In this study, a new quantitative method is described for the determination of five Alternaria toxins and citrinin in tomato and tomato juice samples based on LC-MS/MS detection. Samples were extracted with pure methanol, followed by a derivatisation step with 2,4-dinitrophenylhydrazine to improve the determination of tenuazonic acid and to decrease the wide polarity difference between the compounds of interest. Samples were purified on hydrophilic-modified styrene polymer solid-phase extraction cartridges. High-performance liquid chromatographic columns packed with different core-shell materials were tested for the separation of toxins and a C-18 phase was in the final method applied to achieve sufficient separation of all relevant analytes. A key element of this approach was to prove successful transferability of the method to three different triple quadrupole mass spectrometers. A full single laboratory method validation was performed on two LC-MS/MS systems and performance characteristics met the predefined requirements. Moreover, the method was used in an international proficiency test and the satisfactory z-scores obtained (-0.1 to 0.8 in tomato juice samples) demonstrated the reliability of the approach described. The method will be validated in an inter-laboratory collaborative study and if the criteria for method precision are met, the method will be proposed as a new Work Item to the European Committee for Standardisation.
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Ammonium formate solution, BioUltra, 10 M in H2O
Sigma-Aldrich
Methanol solution, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Hexane, anhydrous, 95%
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Methanol
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Triethylamine, ≥99%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Ammonium formate, ≥99.995% trace metals basis
Sigma-Aldrich
Triethylamine, for amino acid analysis, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Triethylamine, ampule, ≥99.5% (GC)