Merck
  • Cytoplasmic nucleophosmin has elevated T199 phosphorylation upon which G2/M phase progression is dependent.
Cytoplasmic nucleophosmin has elevated T199 phosphorylation upon which G2/M phase progression is dependent.
Scientific reports (2015-07-01)
Narisa Chan, Tit Meng Lim
ABSTRACT
The cytoplasmic mutant of nucleophosmin (NPMc) is found approximately in one-third of acute myeloid leukemia (AML) cases and is highly associated with normal karyotype. Whereas previous studies have focused on wtNPM in centrosome duplication, we further elucidate the role of NPM in the cell cycle by utilizing the increased cytoplasmic load of NPMc. Overexpression of NPMc causes increased phosphorylation of NPM on T199 and, to a lesser degree, S4. T199 phosphorylation is dependent on cdk2 but activators of cdk2 were not elevated. Upon inhibition of cdk2, NPMc-overexpressing cells demonstrate a greater G2/M phase arrest than wtNPM or GFP counterparts. However, the number of cells with 2 centrosomes did not increase concordantly. This suggests that the arrest was caused by a delay in centrosome duplication, most likely due to the inhibition of centrosome duplication caused by unphosphorylated NPMc. Overall, these results suggest that the phosphorylation of T199 is important in the mitotic progression of NPMc-expressing cells. This further supports the hypothesis that NPMc is associated with normal karyotypes in AML because the higher cytoplasmic load of NPM can better suppress centrosome overduplication which would otherwise result in unequal segregation of chromosomes during mitosis, leading to aneuploidy and other genomic instabilities.
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Cdk2
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
MISSION® esiRNA, targeting human CDK2