Merck
  • Sensitive and Selective Determination of Orotic Acid in Biological Specimens Using a Novel Fluorogenic Reaction.
Sensitive and Selective Determination of Orotic Acid in Biological Specimens Using a Novel Fluorogenic Reaction.
Journal of fluorescence (2015-06-01)
Sheng Yin, Shpend Dragusha, Valon Ejupi, Takayuki Shibata, Tsutomu Kabashima, Masaaki Kai
ABSTRACT
Orotic acid is an intermediate in the synthesis pathway of uridine-5'-monophosphate, and increases in body fluids of patients suffering from hereditary disorders such as orotic aciduria and hyperammonemia. In this study, we developed a spectrofluorometric method with or without high-performance liquid chromatography for the selective and sensitive quantification of orotic acid in human biological specimens, using 4-trifluoromethylbenzamidoxime (4-TFMBAO) as a fluorogenic reagent. This reagent provided intensive fluorescence for only orotic acid amongst 62 compounds including structurally related bio-substances such as nucleic acid bases, nucleosides, nucleotides, amino acids, vitamins, bilirubin, uric acid, urea, creatine, creatinine and sugars. Under optimized reaction conditions, orotic acid was reacted with 4-TFMBAO, K3[Fe(CN)6] and K2CO3 in an aqueous solution. The fluorescence produced from the orotic acid derivative was measured at an excitation of 340 nm and an emission of 460 nm. A concentration of 1.2 μM orotic acid per 1.0 mM creatinine in normal urine and 0.64 nmol orotic acid per 5.0 × 10(5) HeLa cells were determined by this method. The present method permitted the facile quantification of orotic acid in healthy human urine and cultured HeLa cells by spectrofluorometry and/or high-performance liquid chromatography.
Sigma-Aldrich
Benzamidoxime, AldrichCPR
Sigma-Aldrich
Potassium carbonate, powder, 99.99% trace metals basis
Sigma-Aldrich
Benzamide oxime, 97%
Sigma-Aldrich
Potassium carbonate, 99.995% trace metals basis
Sigma-Aldrich
Potassium carbonate, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Potassium carbonate, meets USP testing specifications
Sigma-Aldrich
Orotic acid, ≥98% (titration), anhydrous