Merck
CHD1 acts via the Hmgpi pathway to regulate mouse early embryogenesis.
Development (Cambridge, England) (2015-06-21)
Shinnosuke Suzuki, Yusuke Nozawa, Satoshi Tsukamoto, Takehito Kaneko, Ichiro Manabe, Hiroshi Imai, Naojiro Minami
ABSTRACT
The protein CHD1 is a member of the family of ATPase-dependent chromatin remodeling factors. CHD1, which recognizes trimethylated histone H3 lysine 4, has been implicated in transcriptional activation in organisms ranging from yeast to humans. It is required for pre-mRNA maturation, maintenance of mouse embryonic stem cell pluripotency and rapid growth of the mouse epiblast. However, the function(s) of CHD1 in mouse preimplantation embryos has not yet been examined. Here, we show that loss of CHD1 function led to embryonic lethality after implantation. In mouse embryos in which Chd1 was targeted by siRNA microinjection, the expression of the key regulators of cell fate specification Pou5f1 (also known as Oct4), Nanog and Cdx2 was dramatically decreased, starting at mid-preimplantation gene activation (MGA). Moreover, expression of Hmgpi and Klf5, which regulate Pou5f1, Nanog and Cdx2, was also significantly suppressed at zygotic gene activation (ZGA). Suppression of Hmgpi expression in Chd1-knockdown embryos continued until the blastocyst stage, whereas suppression of Klf5 expression was relieved by the morula stage. Next, we rescued HMGPI expression via Hmgpi mRNA microinjection in Chd1-knockdown embryos. Consequently, Pou5f1, Nanog and Cdx2 expression was restored at MGA and live offspring were recovered. These findings indicate that CHD1 plays important roles in mouse early embryogenesis via activation of Hmgpi at ZGA.
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
2-Mercaptoethanol, Molecular Biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
Bovine Serum Albumin, powder, BioXtra
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
Sodium pyruvate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Anti-Nanog Antibody, NT, Chemicon®, from rabbit
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
SAFC
L-Glutamine
Sigma-Aldrich
L-Glutamine