Merck
  • Synthesis, cytotoxicity and antimicrobial activity of thiourea derivatives incorporating 3-(trifluoromethyl)phenyl moiety.
Synthesis, cytotoxicity and antimicrobial activity of thiourea derivatives incorporating 3-(trifluoromethyl)phenyl moiety.
European journal of medicinal chemistry (2015-06-30)
Anna Bielenica, Joanna Stefańska, Karolina Stępień, Agnieszka Napiórkowska, Ewa Augustynowicz-Kopeć, Giuseppina Sanna, Silvia Madeddu, Stefano Boi, Gabriele Giliberti, Małgorzata Wrzosek, Marta Struga
ABSTRACT
A total of 31 of thiourea derivatives was prepared reacting 3-(trifluoromethyl)aniline and commercial aliphatic and aromatic isothiocyanates. The yields varied from 35% to 82%. All compounds were evaluated in vitro for antimicrobial activity. Derivatives 3, 5, 6, 9, 15, 24 and 27 showed the highest inhibition against Gram-positive cocci (standard and hospital strains). The observed MIC values were in the range of 0.25-16 μg/ml. Inhibitory activity of thioureas 5 and 15 against topoisomerase IV isolated from Staphylococcus aureus was studied. Products 5 and 15 effectively inhibited the formation of biofilms of methicillin-resistant and standard strains of Staphylococcus epidermidis. Moreover, all obtained thioureas were evaluated for cytotoxicity and antiviral activity against a large panel of DNA and RNA viruses. Compounds 5, 6, 8-12, 15 resulted cytotoxic against MT-4 cells (CC50 ≤ 10 μM).
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Ethidium bromide solution, BioReagent, Molecular Biology, 500 μg/mL in H2O
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Ethidium bromide solution, BioReagent, Molecular Biology, 10 mg/mL in H2O
Supelco
Phosphorus pentoxide, ≥99.99% trace metals basis
Sigma-Aldrich
Phosphorus pentoxide, puriss. p.a., ACS reagent, ≥98.0% (T)
Sigma-Aldrich
Phosphorus pentoxide, desiccant, with moisture indicator
Sigma-Aldrich
Ethidium bromide solution, suitable for fluorescence, ~1% in H2O
Sigma-Aldrich
Phosphorus pentoxide, powder, ACS reagent, ≥98.0%
Sigma-Aldrich
Phosphorus pentoxide, ReagentPlus®, 99%
Sigma-Aldrich
3-(Trifluoromethyl)aniline, ≥99%
Supelco
Pyridaben, PESTANAL®, analytical standard
Sigma-Aldrich
Phosphorus pentoxide, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethidium bromide, ~95% (HPLC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethidium bromide, BioReagent, Molecular Biology, powder
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis