Merck
  • Investigation of chloromethane complexes of cryptophane-A analogue with butoxy groups using ¹³C NMR in the solid state and solution along with single crystal X-ray diffraction.
Investigation of chloromethane complexes of cryptophane-A analogue with butoxy groups using ¹³C NMR in the solid state and solution along with single crystal X-ray diffraction.
Magnetic resonance in chemistry : MRC (2015-06-23)
Emilie Steiner, Renny Mathew, Iwan Zimmermann, Thierry Brotin, Mattias Edén, Jozef Kowalewski
ABSTRACT
Host-guest complexes between cryptophane-A analogue with butoxy groups (cryptophane-But) and chloromethanes (chloroform, dichloromethane) were investigated in the solid state by means of magic-angle spinning (13)C NMR spectroscopy. The separated local fields method with (13)C-(1)H dipolar recoupling was used to determine the residual dipolar coupling for the guest molecules encaged in the host cavity. In the case of chloroform guest, the residual dipolar interaction was estimated to be about 19 kHz, consistent with a strongly restricted mobility of the guest in the cavity, while no residual interaction was observed for encaged dichloromethane. In order to rationalize this unexpected result, we performed single crystal X-ray diffraction studies, which confirmed that both guest molecules indeed were present inside the cryptophane cavity, with a certain level of disorder. To improve the insight in the dynamics, we performed a (13)C NMR spin-lattice relaxation study for the dichloromethane guest in solution. The system was characterized by chemical exchange, which was slow on the chemical shift time scale but fast with respect to the relaxation rates. Despite these disadvantageous conditions, we demonstrated that the data could be analyzed and that the results were consistent with an isotropic reorientation of dichloromethane within the cryptophane cavity.
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
1,1,2,2-Tetrachloroethane-d2, ≥99.5 atom % D