Merck
  • Simvastatin protects Sertoli cells against cisplatin cytotoxicity through enhanced gap junction intercellular communication.
Simvastatin protects Sertoli cells against cisplatin cytotoxicity through enhanced gap junction intercellular communication.
Oncology reports (2015-08-12)
Lingzhi Wang, Jianxin Peng, Huansen Huang, Qin Wang, Meiling Yu, Liang Tao
ABSTRACT
Cisplatin, an important chemotherapeutic agent against testicular germ cell cancer, induces testicular toxicity on Leydig and Sertoli cells, leading to serious side-effects such as azoospermia and infertility. In a previous study, it was found that simvastatin enhanced the sensitivity of Leydig tumor cells to chemotherapeutic toxicity through the enhancement of gap junction functions. In the present study, the effect of simvastatin on the sensitivity of normal Sertoli cells to cisplatin and the role of gap junctions in such effects was investigated. The results showed that, simvastatin attenuated cisplatin toxicity only when cells exhibited high-density culture where gap junctional formation was possible. When gap junction function was decreased by the gap junction inhibitor or by siRNA targeting connexin 43, the protective effect of simvastatin to cisplatin toxicity was substantially attenuated. Simvastatin also enhanced gap junction functions between Sertoli cells. This effect was mediated by the reduction of PKC-mediated connexin phosphorylation, thereby increasing connexin 43 membrane localization. Thus, simvastatin-induced enhancement of gap junction‑mediated intercellular communication attenuated cisplatin toxicity on Sertoli cells. This result indicated that enhancement of gap junction function by simvastatin may have bilateral beneficial effects on cisplatin‑based chemotherapy, enhancing cisplatin killing on cancer while ameliorating the reproduction toxicity.
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Calcein-AM, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
cis-Diamineplatinum(II) dichloride, ≥99.9% trace metals basis
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Npm1
Sigma-Aldrich
Calcein-AM, Small Package (20 X 50 μg ), ≥90.0% (HPLC)
Sigma-Aldrich
Simvastatin, ≥97% (HPLC), solid
Sigma-Aldrich
Calcein AM solution, 4 mM in DMSO, ≥90% (HPLC), solution
Sigma-Aldrich
cis-Diammineplatinum(II) dichloride, crystalline
Sigma-Aldrich
trans-Platinum(II)diammine dichloride
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Gja1
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Gja3
Sigma-Aldrich
MISSION® esiRNA, targeting human GJA1
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)