Merck
  • Treatment of a mixture of food color additives (E122, E124 and E129) in different water matrices by UVA and solar photoelectro-Fenton.
Treatment of a mixture of food color additives (E122, E124 and E129) in different water matrices by UVA and solar photoelectro-Fenton.
Water research (2015-06-10)
Abdoulaye Thiam, Ignasi Sirés, Enric Brillas
ABSTRACT
The degradation of 130 mL of mixtures of food azo dyes E122, E124 and E129 has been studied by electro-Fenton (EF) and UVA photoelectro-Fenton (PEF) using a stirred tank reactor with either a boron-doped diamond (BDD) or Pt anode and an air-diffusion cathode. The main oxidant was hydroxyl radical formed at the anode from water oxidation and in the bulk from Fenton's reaction between added Fe(2+) and H2O2 generated at the cathode. In sulfate medium, fast decolorization was found for all systems, but the almost total mineralization was more rapidly achieved by PEF with BDD. The performance with a real water matrix was slightly worse, although the removal of total organic load was still as high as 95%. The solar PEF (i.e., SPEF) treatment of dye mixtures using a 2.5 L flow plant with a BDD/air-diffusion cell coupled to a planar solar photoreactor is also reported. Fast decolorization and almost total mineralization was found in the presence of either sulfate, perchlorate, nitrate or a mixture of sulfate + chloride ions. In chloride medium, however, the formation of recalcitrant chloroderivatives decelerated the degradation process. Greater current efficiency and lower specific energy consumption were attained in sulfate medium at lower current density and higher azo dye content. A plausible reaction sequence based on 18 aromatic intermediates identified by GC-MS and 6 short-linear carboxylic acids detected by ion-exclusion HPLC has been proposed. The SPEF process promoted the photodegradation of Fe(III)-oxalate complexes and other undetected products. Sulfate and nitrate ions were always released to the medium.
Sigma-Aldrich
Sodium nitrate, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium sulfate, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium sulfate, ≥99.0%, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium nitrate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium nitrate-14N, 99.95 atom % 14N
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium nitrate, ≥99.0%, suitable for plant cell culture
Sigma-Aldrich
Sodium nitrate, 99.995% trace metals basis
SAFC
Sodium chloride solution, 5 M