Merck
Acquisition of new tumor cell properties by MSC-derived exosomes.
International journal of oncology (2015-05-13)
Yuanyuan Yang, Vesna Bucan, Heike Baehre, Juliane von der Ohe, Anna Otte, Ralf Hass
ABSTRACT
Interaction between multi-functional mesenchymal stroma/stem cells (MSC) and human tumor cells involves the exchange of biological material via extracellular vesicles including exosomes. Protein analysis of MSC-derived exosomes demonstrated the presence of MMP-2 and MSC-specific markers including CD90 and ecto-5'-nucleotidase (CD73). Incubation of tumor cells with these membranous particles revealed a rapid uptake of MSC-released microvesicles whereby breast cancer cells incorporated ~19% and SCCOHT-1 cells representing a rare type of small cell ovarian cancer assimilated ~28% of available exosomes within 24 h. This interaction was accompanied by functional alterations of tumor cell properties during integration of exosomal content from MSC. Indeed, exosome-associated MMP-2 exhibited functional enzyme activity and MCF-7 breast cancer cells with undetectable MMP-2 protein acquired expression of this enzyme and corresponding gelatinase functionality after stimulation with MSC-derived exosomes. Similar effects were observed in SCCOHT-1 cells during culture in the presence of MSC-derived exosomes which enabled new metabolic activities in this tumor cell type. Together, these findings demonstrated that the internalization of MSC-derived exosomes was associated with the acquisition of new tumor cell properties by altering cellular functionalities and providing the capability to re-organize the tumor microenvironment.
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
Adenosine, suitable for cell culture, BioReagent
Sigma-Aldrich
Adenosine, ≥99%
SAFC
L-Glutamine
Sigma-Aldrich
Bromophenol Blue, titration: suitable
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Bromophenol Blue, ACS reagent
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Glycerol, BioUltra, for molecular biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Adenosine
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Hexane, anhydrous, 95%
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Adenosine 5′-monophosphate monohydrate, from yeast, ≥97%
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)